Molecular Biology
Automated Sequencing
Cloning and Expression
Key Resources
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Expression Vectors
Cloning & Expression
For Bacterial, Mammalian and Insect Cell Systems
Featured are a variety of tags, promoters and elements for secretion, transient, stable and bicistronic expression.
  • Our exclusive FLAG® and 3xFLAG® tags are provided in N-terminal, C-terminal, and dual tag configurations in bacterial, mammalian and insect vectors for high quality detection and purification using ANTI-FLAG® antibodies, resins, and 96-well plates.
  • Unique BICEP™ vectors contain an optimized EMCV IRES for high levels of bicistronic expression in mammalian cells for optimal maintenance of stable cell lines and multi-gene expression.
  • The new MAT™ (Metal Affinity Tag) has been incorporated in many of our popular bacterial, mammalian, and insect vectors for high quality downstream purification using HIS-Select™ affinity gels.
  • Sigma's pPolh transfer vectors are the first to offer the well-known FLAG tag for expression of FLAG fusions in insect cell systems.
Bacterial Expression Vectors
Mammalian Expression Vectors
Insect Expression System
Vectors by Affinity Tag
View all Expression Vectors.
back to top
back to Cloning & Expression