Molecular Biology
Automated Sequencing
Cloning and Expression
Key Resources
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 BICEP™ Vectors for Bicistronic Expression
Cloning & Expression
Superior Stable Expression in Mammalian Cells
pBICEP-CMV-1 and pBICEP-CMV-2 offer selection of cells expressing high levels of recombinant protein by expression of a target gene and the neor gene from a single bicistronic message (Figure 1). The neor gene is translated in a cap-independent fashion under control of the EMCV IRES. These vectors lack a mammalian origin of replication and thus likely form stable transfectants through genomic integration of plasmid DNA.
  • Stable expression of target from bicistronic message
  • Expression levels outperform competition >30-fold (Figure 2)
  • Stable cell line production without concern of plasmid loss
  • Maintains high expression levels after numerous passages (Figure 3)
  • FLAG® and 3xFLAG® tags
Product MCS Region
E0779, pBICEP-CMV™-1
N-terminal Met-FLAG
pBICEP-CMV-1
E0904, pBICEP-CMV™-2
N-terminal Met-3xFLAG
pBICEP-CMV-2
back to top
Expression of Two Targets
pBICEP-CMV-3 and pCMV-BICEP-4 are mammalian bicistronic vectors for transient co-expression of any two genes of interest from one construct. They may also be used to create a customized stable selection vector by cloning the gene of interest with the desired selection marker. Target genes are cloned into the two separate MCS regions. Genes cloned into MCS1 are expressed in a cap dependent manner while genes cloned into MCS2 are translated in a cap-independent fashion under control of the EMCV IRES. Because both genes are expressed from one construct, it is likely that most transfected cells will express both genes unlike co-transfected populations. As a result, pBICEP-CMV-3 and pCMV-BICEP-4 are ideal for multisubunit proteins, multi-protein complexes, and protein-protein interaction studies. These vectors contain the SV40 replication origin for episomal replication in appropriate host cells.
  • Transient expression of two targets
  • Most transfected cells will express both targets unlike co-transfected populations
  • Ideal for protein-protein interaction studies
  • Create your own stable selection vector
  • FLAG® and c-Myc tags
Product MCS Region
E7029, pBICEP-CMV™-3
Target 1: N-terminal Met-FLAG
Target 2: No tag
pBICEP-CMV-3
E5905, pBICEP-CMV™-4
Target 1: N-terminal Met-FLAG
Target 2: N-terminal c-Myc
pBICEP-CMV-4
Learn more about BICEP Expression Vectors.
back to top
back to Expression Vectors