Molecular Biology
Automated Sequencing
Cloning and Expression
Key Resources
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Bacterial FLAG® Expression Kits
Cloning & Expression
E. coli Amino-Terminal FLAG Expression Kit (FL-A)
The E. coli Amino-Terminal FLAG Expression Kit provides the specialized FLAG components needed to perform expression of a recombinant N-terminal FLAG fusion in Escherichia coli. The pFLAG-ATS™ (E 5769) and pFLAG-MAC™ (E 5644) Expression Vectors provide a choice of secreted or cytoplasmic expression of N-terminal FLAG fusion proteins, respectively, under control of the i>tac promoter. Also provided in the kit are controls for expression, detection, affinity purification and enterokinase removal of N-terminal FLAG.
Components
  • ANTI-FLAG® M1 monoclonal antibody
  • ANTI-FLAG® M2 affinity gel
  • ANTI-FLAG® M2 monoclonal antibody
  • Enterokinase
  • FLAG® peptide
  • pFLAG-ATS–BAP™ control plasmid
  • pFLAG–ATS™ expression vector (OmpA leader)
  • pFLAG–MAC™ expression vector (no leader)
  • C-24 sequencing primer
  • N-26 sequencing primer
  • N-terminal FLAG–BAP™ control protein
Product Code Description Size
FL-A E. coli Amino-Terminal FLAG® Expression Kit 1 kit
back to top
E. coli Carboxy-Terminal FLAG Expression Kit (FL-C)
The E. coli Carboxy-Terminal FLAG Expression Kit provides the specialized FLAG components needed to perform expression of a recombinant C-terminal FLAG fusion in Escherichia coli. The pFLAG-CTS™ (E 5269) and pFLAG-CTC™ (E 5394) Expression Vectors provide a choice of secreted or cytoplasmic expression of C-terminal FLAG fusion proteins, respectively, under control of the tac promoter. Also provided in the kit are controls for expression, detection, and affinity purification.
Components
  • ANTI-FLAG® M1 monoclonal antibody
  • FLAG® peptide
  • pFLAG-CTC™ expression vector (no leader)
  • pFLAG–CTS-BAP™ control plasmid
  • pFLAG–CTS™ expression vector (OmpA leader)
  • N-26 sequencing primer
  • C-24 sequencing primers
  • C-terminal FLAG–BAP™ control protein
Product Code Description Size
FL-C E. coli Carboxy-Terminal FLAG® Expression Kit 1 kit
back to top
E. coli FLAG-Shift™ Expression Kit (FL-S)
The E. coli FLAG-Shift™ Expression Kit provides the specialized FLAG components needed for expression of an open reading frame (ORF) in all three reading frames as N-terminal FLAG fusion proteins in Escherichia coli. The pFLAG-Shift12™ (E 8023) and pFLAG-Shift12c™ (E 7898) Expression Vectors provide a choice of secreted or cytoplasmic expression of N-terminal FLAG fusion proteins, respectively, under control of the tac promoter. Both vectors contain 12 thymidine residues just upstream from the MCS region that result in frame-shifting during translation. Therefore, simultaneous expression of all three reading frames occurs. This feature is useful when the ORF is unknown as in genomic, cDNA or shotgun libraries. Also provided in the kit are controls for expression, detection, affinity purification and enterokinase removal of N-terminal FLAG.
Components
  • ANTI-FLAG® M1 monoclonal antibody
  • ANTI-FLAG® M2 affinity gel
  • ANTI-FLAG® M2 monoclonal antibody
  • Enterokinase
  • FLAG® peptide
  • FLAG-Shift™12–BAP control plasmid
  • FLAG-Shift™12 expression vector (no leader)
  • FLAG-Shift™12 expression vector (OmpA leader)
  • C-24 sequencing primer
  • N-26 sequencing primer
  • N-terminal FLAG–BAP™ control protein
Product Code Description Size
FL-S E. coli FLAG-Shift™ Expression Kit 1 kit
back to top
back to Expression Vectors