Molecular Biology
Automated Sequencing
Cloning and Expression
Key Resources
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 New MAT™ System Expression Vectors
Cloning & Expression
The new MAT tag or Metal Affinity Tag (HNHRHKH) provides for highly sensitive detection using the Anti-MAT™ antibody and robust one-step purification of recombinant MAT fusion proteins using HIS-Select™ Nickel and Cobalt Affinity Gels.
Bacterial Expression-tac Promoter
  • Cytoplasmic expression
  • N- or C- terminal fusion proteins
  • Ampicillin resistance, T1T2 transcriptional terminator, pMB1(derivative of pBR322) origin of replication, and lacI gene for repression of the tac promoter
  • Expression levels using the tac promoter may be in excess of 10 mg/L of culture when using IPTG as a de-repressor
  • Expression in any established Escherichia coli expression host
Product MCS Region
E5530, pTAC-MAT-1
N-terminal MAT
pTAC-MAT-1
E5405, pTAC-MAT-2
C-terminal MAT
pTAC-MAT-2
back to top
Bacterial Expression-T7 Promoter
  • Cytoplasmic expression
  • N- or C- terminal fusions, dual tag configurations
  • FLAG® vectors for highly sensitive detection and purification using ANTI-FLAG® antibodies, resins, and plates
  • Ampicillin resistance, T1T2 transcriptional terminator, pMB1 (derivative of pBR322) origin of replication, and lacI gene for repression of the T7 promoter
  • Higher yields of recombinant protein than tac system
  • lac operator (lacO) sequences reduce leaky expression
  • Requires hosts with a source of the T7 polymerase such as (DE3) lysogenic strains
T7 Promoter System
Product MCS Region
E5780, pT7-MAT™-1
N-terminal MAT
pT7-MAT-1
E5655, pT7-MAT™-2
C-terminal MAT
pT7-MAT-2
E5280, pT7-FLAG-MAT™-1
N-terminal Met-FLAG, C-terminal MAT (dual tag)
pT7-FLAG-MAT-1
E4905, pT7-MAT™-FLAG-2
N-terminal MAT,C-terminal FLAG (dual tag)
pT7-MAT-FLAG-2
E5155, pT7-MAT-FLAG™-1
N-terminal MAT-FLAG (dual tag)
pT7-MAT-FLAG-1
E5030, pT7-FLAG-MAT™-2
C-terminal FLAG-MAT (dual tag)
pT7-FLAG-MAT-2
back to top
Mammalian Expression
  • Transient, cytoplasmic expression
  • N- or C- terminal dual tag configurations
  • FLAG® vectors for highly sensitive detection and purification using ANTI-FLAG® antibodies, resins, and plates
  • Strong human cytomegalovirus (CMV) promoter regulatory region drives constitutive protein expression levels as high as 1 mg/L in COS cells (in less potent cell lines, protein levels are typically ~0.1 mg/L)
  • pMB1 (derivative of pBR322) origin for replication in bacterial cells
  • Ampicillin resistance selection in bacteria
Product MCS Region
C5864, pCMV-FLAG-MAT™-1
N-terminal Met-FLAG, C-terminal MAT (Dual tagged)
pCMV-FLAG-MAT-1
C5989, pCMV-MAT-FLAG™-1
N-terminal MAT-FLAG (Dual tagged)
pCMV-MAT-FLAG-1
C6114, pCMV-FLAG-MAT™-2
C-terminal FLAG-MAT (Dual tagged)
pCMV-FLAG-MAT-2
back to top
Insect Cell Expression
  • N-terminal and C-terminal configurations
  • Compatible with commercially available ORF 1629 deleted baculoviral DNA
  • Strong polyhedrin (polh) promoter results in high-level expression of target genes during the very late phase of infection
  • AcNPV ORF 603 and ORF1629 regions flank the polh promoter and MCS for homologous recombination with baculoviral DNA and generation of recombinant baculovirus
  • pUC origin of replication and β-lactamase gene (ampr for propagation and selection in bacterial cells
Product MCS Region
T6699, pPolh-MAT™-1
N-terminal MAT
pPolh-MAT-1
T6574, pPolh-MAT™-1
C-terminal MAT
pPolh-MAT-2
back to top
back to Expression Vectors