Molecular Biology
Automated Sequencing
Cloning and Expression
Key Resources
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Mammalian FLAG® Expression Kits
Cloning & Expression
Mammalian Amino-terminal FLAG® Transient Expression Kit (FL-MA)
The Mammalian Amino-Terminal FLAG® Transient Expression Kit provides all of the specialized FLAG components needed to perform expression of a recombinant N-terminal FLAG fusion in mammalian cells. The pFLAG-CMV™-1 (E 7273) and pFLAG-CMV™-2 (E 7398) Expression Vectors allow transient expression of secreted or cytoplasmically expressed fusion proteins. Also provided in the kit are controls for expression, detection, and affinity purification.
Components
  • ANTI-FLAG® M2 monoclonal antibody
  • ANTI-FLAG® M5 monoclonal antibody
  • C-CMV-24 sequencing primer
  • N-CMV-30 sequencing primer
  • FLAG® peptide
  • Met-FLAG-BAP™ control protein
  • pFLAG-CMV™-1-BAP control plasmid
  • pFLAG-CMV™-2-BAP control plasmid
  • pFLAG-CMV™-2 expression vector (no leader)
  • pFLAG-CMV™-1 expression vector (preprotrypsin leader)
Product Code Description Size
FL-MA Mammalian Amino-Terminal FLAG® Transient Expression Kit 1 kit
back to top
Mammalian Amino-terminal FLAG® Stable Expression Kit (FL-MA-S)
The Mammalian Amino-Terminal FLAG Stable Expression Kit provides the specialized FLAG components needed for expression of a recombinant N-terminal FLAG fusion in mammalian cells. The pFLAG-CMV™-3 (E 8770) and pFLAG-CMV™-4 (E 1775) Expression Vectors allow transient or stable expression of secreted or cytoplasmically expressed fusion proteins. Stable expression is accomplished by neomycin selection (G 418). Also provided in the kit are controls for expression, detection, and affinity purification.
Components
  • ANTI-FLAG® M2 monoclonal antibody
  • ANTI-FLAG® M5 monoclonal antibody
  • C-CMV-24 sequencing primer
  • N-CMV-30 sequencing primer
  • FLAG® peptide
  • Met-FLAG-BAP™ control protein
  • pFLAG-CMV™-3-BAP control plasmid
  • pFLAG-CMV™-4-BAP control plasmid
  • pFLAG-CMV™-4 expression vector (no leader)
  • pFLAG-CMV™-3 expression vector (preprotrypsin leader)
Product Code Description Size
FL-MA-S Mammalian Amino-terminal FLAG® Stable Expression Kit 1 kit
back to top
Mammalian Carboxy-terminal FLAG® Transient Expression Kit (FL-MC)
The Mammalian Carboxy-Terminal FLAG® Transient Expression Kit provides the specialized FLAG components needed for transient expression of a recombinant C-terminal FLAG fusion in mammalian cells. The pFLAG-CMV™-5a,b,c Expression Vectors (E 3762) make all reading frames of the MCS available at each restriction site for cloning of an open reading frame. All three vectors lack a translational start signal, so the start signal and any secretion leaders must be inserted along with the gene of interest. Also provided in the kit are controls for expression, detection, and affinity purification.
Components
  • ANTI-FLAG® M2 affinity gel
  • ANTI-FLAG® M2 monoclonal antibody
  • C-CMV-24 sequencing primer
  • N-CMV-30 sequencing primer
  • FLAG® peptide
  • pFLAG-CMV™-5b–BAP control plasmid
  • pFLAG-CMV™-5a expression vector
  • pFLAG-CMV™-5b expression vector
  • pFLAG-CMV™-5c expression vector
  • C-terminal FLAG–BAP™ control protein
Product Code Description Size
FL-MC Mammalian Carboxy-Terminal FLAG® Transient Expression Kit 1 kit
back to top
back to Expression Vectors