Molecular Biology
Automated Sequencing
Cloning and Expression
Key Resources
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 tac Promoter System
Cloning & Expression
tac Vectors for High Level Bacterial Expression
  • Options for cytoplasmic expression or secretion
  • FLAG® vectors for highly sensitive detection and purification using ANTI-FLAG® antibodies, resins, and plates
  • MAT™ vectors for high quality purification using HIS-Select™ resins and 96-well plates
  • N- or C- terminal fusions
  • SHIFT™ vectors for expression in all three reading frames
  • Enterokinase removal of N-terminal FLAG tags
  • Expression kits available
View a Vector Selection Guide.
Product MCS Region
E5769, pFLAG-ATS™
N-terminal FLAG (secreted)
pFLAG-ATS
E5644, pFLAG-MAC™
N-terminal Met-FLAG
pFLAG-MAC
E5269, pFLAG-CTS™
C-terminal FLAG (secreted)
pFLAG-CTS
E5394, pFLAG-CTC™
C-terminal FLAG
pFLAG-CTC
E8023, pFLAG-SHIFT12
N-terminal FLAG fusions in 3 reading frames (secreted)
pFLAG-SHIFT12
E7898, pFLAG-SHIFT12C
N-terminal FLAG fusions in 3 reading frames
pFLAG-SHIFT12C
E5530, pTAC-MAT-1
N-terminal MAT
pTAC-MAT-1
E5405, pTAC-MAT-2
C-terminal MAT
pTAC-MAT-2
tac Vector Features
Bacterial tac promoter-based vectors allow expression, detection and purification of recombinant FLAG and MAT (Metal Affinity Tag) fusions in E. coli. These vectors offer a choice of periplasmic (+ompA) or cytoplasmic expression of amino- or carboxy-terminal tagged fusions. tac expression vectors confer ampicillin resistance for easy selection of positive transformants. Additionally, the vectors contain the T1T2 transcriptional terminator, pMB1(derivative of pBR322) origin of replication, the f1 origin, and the lacI gene for repression of the tac promoter. Vectors utilizing the strong tac promoter (a hybrid of the E. coli trp and lac promoters) offer protein expression levels in excess of 10 mg/L of culture when using IPTG as a de-repressor. These vectors can be used to express protein in any established E. coli expression host.
MAT Tag
The MAT tag or Metal Affinity Tag (HNHRHKH) has been created for purification of recombinant MAT fusion proteins using HIS-Select Nickel and Cobalt Affinity Gels. HIS-Select products allow for highly selective purification of histidine-tagged fusion proteins such as MAT fusions. Many of our newest vectors make use of the MAT tag, often in combination with the well-known FLAG tag. MAT tag containing vectors are offered in formats for N-terminal or C-terminal tagging.
SHIFT Vectors
FLAG-SHIFT vectors are available for sequences where the amino-terminus has not been precisely defined. These vectors contain a "shift" sequence that allows expression of an insert in all three reading frames, thus without regard to the reading frame in which it was originally cloned.
Enterokinase Removal of FLAG Tags
The recognition sequence for enterokinase, Asp-Asp-Asp-Asp-Lys, is found at the C-terminal end of the FLAG epitope tag. Removal of FLAG is possible in all fusion proteins containing an N-terminal FLAG sequence. Dual tag fusion proteins may also be cleaved with enterokinase for removal of one or more tags, depending on the position of FLAG in the protein sequence.
back to top
back to Expression Vectors