Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Amplification Grade DNase I
DNA and RNA Purification
Description: Amplification Grade DNase I (Deoxyribonuclease I) is an endonuclease isolated from bovine pancreas that digests double- and single-stranded DNA into oligo- and mono-nucleotides. The DNase I is suitable for eliminating DNA from RNA preparations prior to sensitive applications, such as RT-PCR. Since no RNA purification procedure removes 100% of the DNA, RNA samples should be digested with DNase I before RT-PCR. A simple 15 minute digestion at room temperature removes the contaminating DNA. The DNase I is inactivated by adding the stop solution provided and heating. Heating also denatures the RNA, so the RNA can be used directly for reverse transcription.
Many commercial DNase I formulations are contaminated with residual RNases. This RNase contamination can destroy or degrade valuable RNA samples prior to reverse transcription. Laboratory comparisons have shown that Sigma's Amplification Grade DNase I demonstrates lower RNase activity than that from several leading molecular biology product suppliers.
Features and Benefits
Convenient
Eliminates DNA from RNA preparations
Return to Top
Amplification Grade DNase I
Product Number
Product Description
Quantity
Amplification Grade DNase I (Sufficient for treating at least
1,000 µg of RNA)
1 Kit
Return to Top
Kit Contents
  • Amplification Grade DNase I (1,000 units)
  • 10X reaction buffer
  • Stop Solution
Return to Top
Sigma's Amplification Grade DNase I has the lowest RNase activity
Agarose gel of lowest RNase Activity
Figure 1. A 1 µg aliquot of a 1.9 kb in vitro transcription product was incubated with 1 unit of each DNase I at 37°C for 1 hour and analyzed on a 1% agarose gel. Cu = unincubated control (RNA in buffer without DNase, kept on ice).
Ci = incubated control (RNA in buffer but without DNase, incubated at 37°C for 1 hour).
Note: To determine the effectiveness of DNase I treatment, parallel PCR reaction should be run without adding reverse transcriptase to check for amplification from contaminating DNA.
Return to Top
RT-PCR sensitivity with or without DNase digest
RT-PCR Sensitivity
Figure 2. RNA was prepared from HeLa cells with the GenElute Mammalian Total RNA Kit (RTN). A 1000, 100, 10, 1 or 0.1 ng aliquot of RNA was digested with Amplification Grade DNase I and amplified by RT-PCR. * Indicates reactions without reverse transcriptase.
Note: Without DNase treatment, a PCR product is obtained without reverse transcriptase, indicating that the RNA is contaminated with genomic DNA. DNase treatment eliminated this PCR-only product. RT-PCR products are visible down to 0.1 ng RNA with or without DNase, demonstrating no loss of sensitivity with the DNase treatment.
Return to Top