Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Extract-N-Amp™ Reagents
DNA & RNA Purification
Get more from your Extract-N-Amp™ PCR Kit! Sigma offers Extract-N-Amp supplementary reagents to use in conjunction with our Extract-N-Amp kits. The PCR ReadyMixes allow increased amplifications from the Extract-N-Amp PCR Kits, and accommodate dilution experiments prior to PCR amplification.
Sigma Extract-N-Amp PCR Kits offer an innovative extraction system that prepares genomic DNA for amplification by PCR or qPCR in as little as 8 minutes. The optimized kits include a specially formulated hot start PCR ReadyMix™ for amplification directly from an aliquot of the stable extract. Suitable for most downstream applications, our Extract-N-Amp kits provide the perfect solution for genomic DNA analysis.
Watch the video protocol
Rapid Genotyping of Mouse Tissue Using Sigma's Extract-N-Amp Tissue PCR Kit
Video brought to you by JoVE.com
Re-amplification mixes for Extract-N-Amp Tissue and Plant Kits
Product Code Name Package
Size
Add to Cart
E3004 Extract-N-Amp PCR ReadyMix™ 1.2 ml
12 ml
125 ml
R4775 REDExtract-N-Amp PCR ReadyMix 1.2 ml
12 ml
125 ml
Re-amplification mixes for Extract-N-Amp Blood Kits
Product Code Name Package
Size
Add to Cart
P8115 Extract-N-Amp PCR ReadyMix for Blood 12 ml
P8240 Extract-N-Amp PCR ReadyMix for Blood 12 ml
Supplementary PCR diluent for Extract-N-Amp Tissue, Plant and Seed Kits
Product Code Name Package
Size
Add to Cart
E8115 Extract-N-Amp PCR Diluent 100 ml
E8115 Extract-N-Amp PCR Diluent 500 ml
back to top
DNA & RNA Purification home page