Life Science
Antibody Explorer
Automation
Cancer Research
Cell Culture
Cell Signaling and Neuroscience
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Nutrition Research
Obesity Research
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Your Favorite Gene - Search
Life Science Quarterly
PathFinder
 Extract-N-Amp Purification Kits
DNA and RNA Purification
Your shortcut for PCR.
Extract-N-Amp™ PCR kits are the world’s first integrated extraction and amplification process for rapid blood, tissue or plant assays. Eliminate the need for any columns or long enzymatic sample purification with this simple “lyse & go” method for PCR. Kits include all reagents, proprietary buffers, plus enzymes needed to rapidly extract and amplify targets of interest from a variety of cells and tissues.
  • Integrated purification and amplification
  • Simplified sample processing
Origami Mouse Graphic
The kits provide all the reagents necessary to rapidly extract and amplify DNA targets of interest by PCR. A novel extraction method eliminates the need for long enzymatic digestions or homogenization. The kit also includes a specially formulated hot start PCR ReadyMix™ for amplification directly from the extract. The PCR ReadyMix comes in two formulations: Extract-N-Amp PCR ReadyMix and REDExtract-N-Amp PCR ReadyMix. The REDExtract-N-Amp PCR ReadyMix contains and inert dye that acts as a tracking dye and allows for convenient loading of PCR reaction onto agarose gels for analysis.
Sigma provides validated protocols to extract and amplify genomic DNA from animal and plant samples.
In a typical procedure, genomic DNA is extracted from a sample that has been incubated in preparation solution and extraction solution at room temperature. The sample is heated and then mixed with a third solution to neutralize inhibitory substances prior to PCR. A portion of the DNA extract is then added to a PCR reaction containing primers and the ReadyMix included in the kit. The remainder of the DNA can be refrigerated for convenient storage.
Typical Applications include:
  • Genotyping
  • Transgene detection
  • Knockout analysis
  • Sequencing
Product Speed Product Description Starting Material
Extract-N-Amp™ Tissue PCR Kit
Extract-N-Amp™ REDExtract-N-Amp Tissue PCR Kit
Less than 15 minutes from extraction to amplification Rapid amplification of genomic DNA from
• Mouse-tails
• Hair
• Animal Tissue
• Buccal Swabs
Tail: 0.5 to 1.0 cm
Hair: 1 root
Tissue: 2 to 10 mg
Buccal: 10 µl
SYBR® Green Extract-N-Amp™ Tissue PCR Kit Less than 15 minutes from extraction to amplification Rapid quantitative PCR of genomic DNA from
• Mouse-tails
• Hair
• Animal Tissue
• Buccal Swabs
Tail: 0.5 to 1.0 cm
Hair: 1 root
Tissue: 2 to 10 mg
Buccal: 10 µl
Extract-N-Amp™ Blood PCR Kit
REDExtract-N-Amp Blood PCR Kit
8 minutes from extraction to amplification Rapid amplification of genomic DNA from
• Whole blood
Blood: 10 µl
Extract-N-Amp™ Plant PCR Kit
REDExtract-N-Amp Plant PCR Kit
Less than 15 minutes from extraction to amplification Rapid extraction and PCR amplification of genomic DNA from
• Plant leaves
Plant Leaf Disk:
0.5 to 0.7 cm
Extract-N-Amp™ Seed PCR Kit
REDExtract-N-Amp Seed PCR Kit
Less than 15 minutes from extraction to amplification Rapid amplification of genomic DNA from
• Plant seeds
1 seed