Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Extract-N-Amp™ Plant PCR Kits
DNA and RNA Purification
Description: The Extract-N-Amp™ Plant PCR Kits contain all the reagents necessary to rapidly extract genomic DNA from plant leaves and amplify targets of interest by PCR.
  • Novel – single-step extraction of genomic DNA to PCR
  • Fast – tissue to PCR in 15 minutes
  • Convenient – no long enzymatic digestions
The Extract-N-Amp Plant PCR Kit offers a novel Extraction Solution that eliminates the need for freezing of plant tissues with liquid nitrogen, mechanical disruption, organic extraction, column DNA purification, or alcohol precipitation. The product includes a specially formulated hot start PCR ReadyMix for amplification directly from the extract. Extract-N-Amp products are Sigma Advanced Technology certified.
Procedure: Genomic DNA is extracted from 0.5 to 0.7 cm plant leaf disks that have been cut with a standard paper punch and simply incubated in Extraction Solution at 95 °C for 10 minutes. An equal volume of Dilution Solution is added to the extract to neutralize inhibitory substances prior to PCR. A portion of the DNA extract is then added directly to the optimized PCR mix supplied.
Application: Perfect for genotyping.
Extract-N-Amp Plant Flow Chart
Beneficial Features
Starting Material
0.5 to 0.7 cm plant leaf disk
Speed
Extraction of genomic DNA for PCR in 15 minutes
Flexible
Extract and amplify target DNA sequences from a variety of plant species (Figure 1)
Specific
Hot Start antibody for highly specific PCR amplification of genomic DNA
Safe
No phenol/chloroform or other hazardous material
Return to Top
Pick the formulation that is right for you.
The Extract-N-Amp Tissue PCR Kits include a specially formulated hot start PCR ReadyMix for amplification directly from the extract.
The PCR ReadyMix comes in two formulations:
  • REDExtract-N-Amp PCR Ready Mix – contains an inert dye that acts as a tracking dye and allows for convenient loading of PCR reactions onto agarose gels for analysis.
  • Extract-N-Amp PCR Ready Mix – no dye contained.
Return to Top
Enter the Origami Photograph Contest - click here
Order Information
Extract-N-Amp Plant PCR Kits
Product Code Name Number of Extractions Number of Amplifications Technical Manual .pdf
XNAP2 Extract-N-Amp Plant PCR Kit 100 100 X
XNAP2E Extract-N-Amp Plant PCR Kit 100 500 X
XNAR Extract-N-Amp Plant PCR Kit 1000 1000 X
XNAP2RE Extract-N-Amp Plant PCR Kit 1000 5000 X
REDExtract-N-Amp Plant PCR Kits
Product Code Name Number of Extractions Number of Amplifications Technical Manual .pdf
XNAPS REDExtract-N-Amp Plant PCR Kit
(contains REDTaq)
10 10 X
XNAP REDExtract-N-Amp Plant PCR Kit
(contains REDTaq)
100 100 X
XNAPE REDExtract-N-Amp Plant PCR Kit
(contains REDTaq)
100 500 X
XNAPR REDExtract-N-Amp Plant PCR Kit
(contains REDTaq)
1000 1000 X
XNAPRE REDExtract-N-Amp Plant PCR Kit
(contains REDTaq)
1000 5000 X
Return to Top
Kit Contents
Extract-N-Amp Kit
  • Extraction Solution
  • Dilution Solution
  • Extract-N-Amp PCR Ready Mix
  • 2 ml Tubes for Extraction (not included with the 1,000 Amplification kit)
REDExtract-N-Amp Kit
  • Extraction Solution
  • Dilution Solution
  • REDExtract-N-Amp PCR Ready Mix
  • 2 ml Tubes for Extraction
Return to Top
Sample Data
PCR analysis of genomic DNA extracted from 5 different plant species using Sigma's Extract-N-Amp Plant Kit.
Sample Data
Figure 1. Genomic DNA was extracted from 0.5 cm leaf disks that were cut using a standard paper punch. DNA was extracted using the Extract-N-Amp Plant PCR Kit in less than 15 minutes. All samples were then amplified using the specially formulated Hot Start PCR mix. The products were generated from a 30-cycle duplex reaction containing primers specific to plant chloroplast (upper band) and primers specific to Cannabis sativa DNA (lower band). MW ladder is 100, 200, 400 and 800 bp. Data provided by Andy Hopwood, Forensic Science Service, Birmingham, England.
Return to Top
Sequence was resolved on an ABI 310 from a purified, 645 bp corn leaf PCR product.
Figure 2. The PCR product was purified with the GenElute™ PCR Clean-Up Kit (Product Code NA1020). The DNA extraction and PCR were performed using Sigma's Extract-N-Amp Plant PCR Kit. The sequence was obtained using the same primers as for the original PCR.
Click Image to View Sequence in its Entirety
Return to Top
Stability of Extract-N-Amp Plant Extracts.
Extract-N-Amp Plant Stability Data
Figure 3. Eight disks were punched from a corn leaf, and DNA was extracted according to the procedure in the Technical Bulletin for the Extract-N-Amp Plant Kit. Two 4 µl aliquots from each were analyzed immediately by quantitative PCR with SYBR® Green detection on an ABI Prism 7700. DNA standards for quantitative PCR were purified DNA prepared from corn leaf tissue with the GenElute Plant Genomic DNA Kit (Product Code G2N70). Half of the leaf extracts were stored at 4 °C (recommended storage conditions) and the other half at 37 °C (accelerated storage). Quantitative PCR was repeated after 1 week, 3 weeks, 6 weeks and 6 months from extracts at 37 °C. Results for storage at 37 °C are shown. The average of 2 replicate PCR assays from each extract is plotted. Error bars represent one standard deviation.
Return to Top
Return to Automation