Life Science
Antibody Explorer
Automation
Cancer Research
Cell Culture
Cell Signaling and Neuroscience
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Nutrition Research
Obesity Research
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Your Favorite Gene - Search
Life Science Quarterly
PathFinder
 GenElute™ Five-Minute Plasmid Miniprep Kit
DNA and RNA Purification
Description: The GenElute™ Five-Minute Miniprep Kit features an ultra streamlined protocol yielding up to 5 µg high-quality plasmid DNA in about five minutes.
  • Sophisticated – no pelleting cells or clearing lysates
  • Flexible – binding column works with any standard laboratory vacuum manifold
  • Plentiful – up to 5 µg of plasmid DNA or enough for 15 sequencing reactions
Scientists at Sigma developed a novel lysis procedure that eliminates pelleting cells, removing media and clearing neutralized lysates. This discovery enables super-rapid purification of plasmid DNA, and qualifies the product for the Sigma Advanced Technology* designation.
Procedure: 400 µl of culture is added to a 2 ml collection tube. Cells are lysed for 1-2 minutes. A Binding Solution is added and the mixture is passed through a binding column. After washing the column to remove trace contaminants, the plasmid DNA is eluted in 10mM Tris HCl, pH 8.5. Molecular biology water may be substituted as an elution reagent.
Application: The DNA is suitable for immediate use in capillary DNA sequencing, clone screening, restriction digestion and PCR.
Beneficial Features
  • Use only 400 µl overnight bacterial culture
  • Purify up to 5 µg from high-copy plasmid
  • Only 5 minutes from start to finish
  • Choose vacuum or spin protocols
Order Information
Product Product Description Technical
Bulletin
Preps
PFM10 GenElute Five-Minute Plasmid Miniprep Kit 10
PFM50 GenElute Five-Minute Plasmid Miniprep Kit 50
PFM250 GenElute Five-Minute Plasmid Miniprep Kit 250
Return to Top
Sample Data
Purified Plasmid DNA is Especially Suited for Automated Sequencing
Figure 1. Capillary DNA sequencing of plasmid DNA purified with the GenElute Five-Minute Plasmid Miniprep Kit. pCMV-SPORT-βgal (7.9 kb) was prepared from 400 µl overnight culture of E. coli host strain DH5α with the GenElute Five-Minute Plasmid Miniprep Kit. A 5 µl sample of the preparation was submitted, with adjustment in DNA concentration, to SeqWright DNA Technology Service (Houston, TX) for sequencing with a T7 sequencing primer and ABI BigDye™ terminator v3.1 on an ABI Prism® 3730 DNA Analyzer. The Phred > 20 score was 905 bases.
  Click Image to View Sequence in its Entirety
Return to Top
Achieve Consistent Yields and Quality for Common Applications
Figure 2. Restriction digestion analysis of pCMV-SPORT-βgal DNA purified with GenElute Five-Minute Plasmid Miniprep Kit. Lanes 1 & 19: 1 kb DNA ladder (Sigma D 0428); lanes 2 to 17: 16 independent plasmid mini preparations; lane 18: uncut pCMV-SPORT-βgal DNA. Each digestion was carried out in 15 µl at 37 °C for 1 hour, with 3 µl of eluate and 3 units each of Eco RI and Xho I. A quarter of each digestion was fractionated in 1% agarose gel.   agarose gel image for 5 minute miniprep
*Sigma Advanced Technology™ distinguishes the scientific team and the product it developed for the achievement of a new technology, a novel invention or measurable improvements resulting in absolute performance advantages over current methods.
Return to Top