Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 GenElute™ Bacterial Genomic DNA Miniprep Kit
DNA and RNA Purification
For purification of genomic DNA from a variety of cultured bacteria
Description: Sigma’s GenElute Bacterial Genomic Kit provides a simple and convenient technique to isolate high quality DNA from both Gram negative (Figure 1) and Gram positive bacteria (Figure 2). This kit combines the advantages of a silica-based system with a microspin format, eliminating the need for expensive resins and hazardous organic compounds. Bacteria are first incubated with the appropriate enzymes to ensure efficient cell lysis and DNA release from the cells.
Quick look: NA2100, NA2110, NA2120
Procedure: Bacteria are first incubated with the appropriate enzymes to ensure efficient cell lysis and DNA release from the cells. The bacteria are then lysed in a chaotropic salt-containing solution. DNA is bound to the silica-based membrane and the remaining lysate is removed by centrifugation. After washing to remove contaminants, the DNA is eluted with buffer into a collection tube. Eluted DNA can be up to 50 kb in length (Figure 3) and is suitable for downstream applications such as restriction endonuclease digestions, PCR, and Southern blots.
GenElute Mammalian Genomic Flow Chart
The GenElute Bacterial Genomic DNA Kit contains all of the reagents needed to purify genomic DNA from Gram negative bacteria (Figure 3). However lysozyme (Product No. L7651), which must be purchased separately, is needed for most Gram positive bacteria to thoroughly lyse the thick peptidoglycan cell walls. A Gram positive Lysis Solution is provided with each GenElute Kit as a diluent for preparing the lysozyme stock solution.
Features and Benefits:
  • Protocols provided for Gram positive and Gram negative bacteria
  • Purified DNA has an A260/A280ratio between 1.6 and 1.9
  • Typical DNA yields of 15 µg-20 µg (See Table 1)
  • Lysozyme diluent & RNase A Solution provided for added convenience
Storage: Room Temperature
R: 22-36/37/38-42 S: 26-36
Application: Purified DNA is suitable for downstream applications such as Restriction Endonuclease Digestions, PCR, and Southern Blots.
Sample Data
Typical DNA Yields with the GenElute Bacterial Genomic DNA Kit.
Source Type of Media Amount of Overnight Culture OD600 per ml Overnight Culture* Typical DNA Yield (with RNase Treatment)**
Escherichia coli,
ATCC# 11775
Terrific broth
(Product Code T9179)
0.8 ml 12.5 20 µg
Escherichia coli,
ATCC# 11775
LB broth
(Product Code L7658)
1.5 ml 5 20 µg
Escherichia coli,
DH10B
LB broth
(Product Code L7658)
1.0 ml 5 15 µg
Pseudomonas fluorescens,
ATCC# 13525
Terrific broth
(Product Code T9179)
0.8 ml 16 25 µg
Pseudomonas fluorescens,
ATCC# 13525
Nutrient broth
(Product Code N7519)
1.5 ml 2 20 µg
Bacillus subtilis,
ATCC# 6051
Todd Hewitt broth
(Product Code T1438)
1.5 ml 6 25 µg
Streptococcus mutans,
ATCC# 35668
Todd Hewitt broth
(Product Code T1438)
1.5 ml 1.3 15 µg***
Staphylococcus epidermidis,
ATCC# 14990
Nutrient broth
(Product Code N7519)
1.5 ml 2 8 µg****
* Values adjusted for dilution factor.
** Based on performing two 200 µl elutions.
*** Lysozyme Solution was supplemented with 250 units/ml of Mutanolysin (Product Code M9901).
**** Lysozyme Solution was supplemented with 200 units/ml of Lysostaphin (Product Code L7386).
Return to Top
Comparison of Gram(–) bacteria genomic DNA isolation kits.
GenElute Bacterial Gram Negative Gel
Q = Supplier Q
S = Sigma
C = Supplier C
Figure 1. Agarose gel analysis of genomic DNA isolated from the indicated Gram negative bacteria prepared using the GenElute Bacterial Genomic DNA Kit versus kits from other suppliers. Equal proportions of DNA were resolved on a 1%, 1X TBE agarose gel. The Lambda Hind III ladder (Product Code D9780) was used as a size standard (M).
Return to Top
Comparison of Gram(+) bacteria genomic DNA isolation kits.
GenElute Bacterial Gram Positive
Q = Supplier Q
S = Sigma
C = Supplier C
Figure 2. Agarose gel analysis of genomic DNA isolated from the indicated Gram(+) bacteria prepared using the GenElute Bacterial Genomic DNA Kit versus kits from other suppliers. Equal proportions of DNA were resolved on a 1%, 1X TBE agarose gel. The Lambda Hind III ladder (Product Code D9780) was used as a size standard.
Return to Top
Pulse Field Gel Electrophoresis (PFGE) of bacterial DNA.
GenElute Bacterial Genomic PFGE
Figure 3. Purified genomic DNA was isolated from various bacterial species using the GenElute Bacterial Genomic DNA kit. A 1 µg aliquot of DNA from each respective bacterial sample was resolved on a 1% agarose gel in 0.5X TBE at 150 volts for 16 hours using a BioRad CHEF DRII system. The initial pulse time was 2 seconds, the final pulse time was 13 seconds, the start ratio was 1.0, pump speed was set at 70, and PFGE was carried out at 4°C. Marker (M) represents the 0.1-200 kb Pulse marker (Product Code D2291).
Return to Top
Lysozyme may dissolve more readily by pipetting the mixture up and down as opposed to vortexing. This also reduces foaming that may
occur from vortexing.
Order Information
Product Code Name Purifications
per kit
Technical
Manual .pdf
Short
Protocol .pdf
NA2100 GenElute Bacterial Genomic DNA Kit 10 X X
NA2110 GenElute Bacterial Genomic DNA Kit 70 X X
NA2120 GenElute Bacterial Genomic DNA Kit 350 X X
Return to Top