Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 GenElute™ HP Plasmid Kits
DNA and RNA Purification
Description: The GenElute™ HP Plasmid Midiprep and Maxiprep Kits offer purification of high-quality plasmid DNA in less than 30 minutes (Figure 1). GenElute HP Plasmid Kits include a filter syringe for lysate clearing, binding columns that can be used with a vacuum or spin purification format, and does not require alcohol precipitation.
  • Fast – high-quality plasmid DNA in 30 minutes
  • Convenient – vacuum or spin column format
  • Cost effective – more value for the money
Sigma Advanced Technology distinguishes the scientific team, and the product it developed, for the achievement of a new technology, a novel invention or measurable improvements resulting in absolute performance advantages over current methods.
The GenElute HP Plasmid preps deliver significant time-savings and overall higher yields compared to other methods and are Sigma Advanced Technology™ certified.
Quick Look: NA0150S, NA0150, NA0160, NA0200S, NA0200, NA0300S, NA0300, NA0310
Procedure: An overnight recombinant E. coli culture is harvested by centrifugation and subjected to a modified alkaline-SDS lysis procedure. The lysate is cleared with a filter syringe and DNA is bound to the silica-based membrane. The remaining contaminates are removed by a wash step. Finally, the bound DNA is eluted in buffer or water.
Application: High-quality DNA is ready for immediate use for the most demanding applications, including transfection (Figure 3), restriction digestion (Figure 4), ligation, automated sequencing (Figure 5).
Beneficial Features
Culture Volume   1-5 ml (Mini), 50 ml (Midi) and 150 ml (Maxi) bacterial culture
Yield   Up to 25 µg (Mini), 350 µg (Midi) and 1.2 mg (Maxi) yield of high-copy
  plasmid DNA (Figure 2)
Speed   From harvested bacterial culture to pure plasmid DNA in 30 minutes or less
Flexible   Offers the flexibility of a vacuum or spin format; does not rely on gravity flow
  or ultracentrifugation
Economic   Lower cost per prep plus higher yields per prep equals double savings
Return to Top
Order Information
GenElute HP Plasmid DNA Kits
Product No. Product Name Purifications
per kit
Technical Manual .pdf Short
Protocol .pdf
NA0150S GenElute HP Plasmid Miniprep Kit 10 X -
NA0150 GenElute HP Plasmid Miniprep Kit 70 X -
NA0160 GenElute HP Plasmid Miniprep Kit 350 X -
NA0200S GenElute HP Plasmid Midiprep Kit 4 X X
NA0200 GenElute HP Plasmid Midiprep Kit 25 X X
NA0300S GenElute HP Plasmid Maxiprep Kit 4 X X
NA0300 GenElute HP Plasmid Maxiprep Kit 10 X X
NA0310 GenElute HP Plasmid Maxiprep Kit 25 X X
Return to Top
Sample Data
Preparation Time and Plasmid DNA Yield for different Maxiprep Kits
Graph for Preparation Time
Figure 1. Comparison of time required per isolation using different Maxiprep plasmid isolation systems. Plasmid DNA was isolated following each manufacturers recommended protocol. Each system was tested multiple times and the average preparation time is shown here.
  Graph for Plasmid DNA Yield
Figure 2. Comparison of plasmid DNA yield using different Maxiprep plasimd isolation systems. Plasmid DNA was isolated following each manufacturers recommended protocol. All samples were done in duplicate.
Return to Top
High-quality DNA for transfections
Graph for transfection of cells
Figure 3. Comparison of transfection efficiencies of plasmids prepared with different plasmid isolation methods. Data shows an average of 3 replicates for each named isolation method. All transfections were in CHO-K1 cells. The OD measurements were taken at 420 nm recording the units of ß-gall/plate. The cells were grown to 60-70% confluency and transfected using 3 µg of plasmid pSPORT-CMV/15 µg of Escort IV (Product No. L3287). b-Gal activity was determined 54 hours post-transfection.
  Restriction Enzyme Results
Figure 4. Restriction digestion of plasmid DNA isolated using the GenElute HP Plasmid Maxiprep Kits and a comparable kit from Supplier Q. The plasmid DNA (800 ng) was digested with 10 units of Hind III at 37 °C for 4 hours. Each plasmid sample represents 100 ng of undigested DNA (left) and digested DNA (right) loaded in each pair of lanes. Samples were loaded on a 1% TBE agarose gel. The marker (M) used was 1 kb DNA Ladder (Product No. D0428).
Return to Top
High-quality DNA for Automated Sequencing
Figure 5. Electropherogram revealing >600 bases of sequence from pCMV-SPORT-b-gal purified from GenElute HP Plasmid Midiprep Kit. Cycle Sequencing was performed using 500 ng template, a T7 sequencing primer and ABI BigDye™ terminator chemistry. Sequencing reactions were resolved on an ABI Prism XL instrument with a 48 cm gel cassette containing 4.5% AutoPAGE Plus acrylamide at 2.4 kV, 48°C for 7 hours.
Click Image to View Sequence in its Entirety
Return to Top