Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
The GenElute™ HP Plasmid Midiprep and Maxiprep Kits offer purification of high quality plasmid DNA in less than 30 minutes (Figure 1). GenElute™ HP Plasmid Kits feature a filter syringe for lysate clearing, and binding columns that can be used with a vacuum or spin purification format.
An overnight recombinant E. coli culture is harvested by centrifugation and subjected to a modified alkaline-SDS lysis procedure. The lysate is cleared with a filter syringe and DNA is bound to the silica-based membrane. The remaining contaminates are removed by a wash step. Finally, the bound DNA is eluted in buffer or water. High quality DNA is suitable for the most demanding applications including transfection (Figure 3), restriction digestion, ligation, automated sequencing.
Features and Benefits
  • From harvested bacterial culture to pure plasmid DNA in 30 minutes or less
  • Up to 350 µg (Midi) and 1.2 mg (Maxi) yield of high-copy plasmid DNA (Figure 2)
  • Offers the flexibility of a vacuum or spin format
Purified Plasmid DNA
Actual Time: Under 30 minutes
1) Resuspend and
Lyse cells
3-5 minutes
2) Neutralize in
filter syringe
5 minutes
3) Prepare Column
Add solution and vacuum
4) Clear lysate
and bind DNA
Filter lysate directly
onto column and
apply vacuum -
5 minutes
5) Wash
Add wash solution
and allow column
to dry on vacuum -
10 minutes
6) Elute
Centrifugation -
5 minutes
LS Registration