Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 GenElute™ PCR Clean-Up
DNA and RNA Purification
Description: The GenElute™ PCR Clean-Up Kit is designed for rapid purification of single-stranded or double-stranded PCR amplification products (100 bp to 10 kb) from other components in the reactions, such as excess primers, nucleotides, DNA polymerase, oil and salts (Figure 1).
Procedure: DNA is bound on a silica membrane within the spin column. The bound DNA is washed and the clean, concentrated DNA is eluted in the buffer of choice. Each column can purify up to 100 µl or 10 µg of PCR amplified DNA and recover up to 95% of PCR products between 100 bp and 10 kb. More than 99% of the primers and most primer-dimers (<40 bp) are removed.
Application: Purified DNA can be used in enzymatic reactions, conventional or Automated Sequencing (Figure 2), and Cloning and Microarray Analysis.
Purification Chart
Beneficial Features
Starting Amount
100 µL or 10 µg of DNA
Yield
Recovers up to 95% of PCR fragments between 100 bp to 10 kb.
Speed
Purify up to 100 µL or 10 µg of PCR amplified DNA in 5 minutes.
Flexible
DNA concentrated in 50 µL of buffer
Economic
40% more purifications than market leader
Return to Top
Order Information
Product Number Name Technical Manual .pdf
NA1020 GenElute PCR DNA Purification Kit (Sufficient for 70 purifications) X
Return to Top
Sample Data
Comparison of PCR product recovery and primer removal.
Purification Gel
Figure 1. Three separate PCR products were purified with GenElute PCR Clean-Up Kit and the kit from Supplier Q. Products were 143 bp from corn leaf, 375 bp from pBR322, 2 kb from human blood. Samples were analyzed on a 20% TBE acrylamide gel and visualized by staining with SyberGreen II™.
Lanes 1: Unpurified Reaction
Lanes 2: GenElute PCR Clean-Up Kit
Lanes 3: Supplier Q
Return to Top
Purified PCR products are suitable for Automated Sequencing.
Figure 2. Sequence was resolved on an ABI 310 from a purified, 645 bp corn leaf PCR product. The PCR product was purified with the GenElute PCR Clean-Up Kit. The sequence was obtained by using ABI BigDye™ Terminator Chemistry and the same primers as for the original PCR.
Click Image to View Sequence in its Entirety
Return to Top