Life Science
Antibody Explorer
Automation
Cancer Research
Cell Culture
Cell Signaling and Neuroscience
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Nutrition Research
Obesity Research
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Your Favorite Gene - Search
Life Science Quarterly
PathFinder
 GenElute™ Plant Genomic DNA Miniprep Kit
DNA and RNA Purification
For purification of genomic DNA from a variety of plant species
Description: With the GenElute Plant Genomic DNA Miniprep Kit, high quality genomic DNA can be purified from a variety of plant species (Table 1, Figure 1, and Figure 2). The kit contains all the reagents, columns and tubes necessary to isolate genomic DNA from up to 100 mg of fresh or 20 mg of freeze-dried plant tissue.
Quick look: G2N10, G2N70, G2N350
Procedure: Plant tissue is disrupted by grinding in liquid nitrogen, and DNA is released with detergent and chaotrope. Proteins, polysaccharides, and cell debris are eliminated with a 10 minute precipitation procedure followed by centrifugation through a filtration column, included in the kit. The genomic DNA is purified further by a silica bind-wash-elute procedure in microcentrifuge spin columns. Purified DNA is ready for downstream applications such as PCR (Figure 2), restriction endonuclease digestions, cloning and southern blots.
Features and Benefits:
  • Less than 40 minutes from tissue to purified genomic DNA, including disruption in liquid nitrogen
  • Typical DNA yields of up to 20 µg per prep
  • No RNase treatment required
  • 40% more purifications per kit than the leading supplier
Storage: Room Temperature
R: 20/21/22-32-36/38 S: 26-36
Application: Purified DNA (Figure 2) is ready for downstream applications such as PCR (Figure 3), restriction endonuclease digestions, cloning and southern blots.
GenElute Mammalian Genomic Flow Chart
Return to Top
Sample Data
Table 1. Typical yields of genomic DNA isolated from various plant species per 100 mg of starting leaf tissue
Material Typical Yield
Corn 7.5 µg
Dianthus tissue culture 3.3 µg
Pepper 3.1 µg
Rice 5.9 µg
Soybean 5.7 µg
Tobacco 5.2 µg
Tomato 6.2 µg
Tomato (20 mg freeze dried leaf tissue) 5.7 µg
Wheat 11.5 µg
Return to Top
Genomic DNA from various plant species isolated with GenElute Plant Genomic DNA Miniprep Kit.   PCR amplification of a 500 bp product isolated from genomic DNA.
Genomic DNA from various plant species isolated with GenElute Plant Genomic DNA Miniprep Kit
Figure 1. Purified genomic DNA (0.4 µg/lane) was analyzed on a 0.8% agarose gel. Markers are Lambda EcoR I (Product No. D9281) Hind III digest.
  PCR amplification of a 500 bp product isolated from genomic DNA
Figure 2. Genomic DNA from soybean leaves was purified using the GenElute™ Plant Genomic DNA Miniprep Kit. A 5 µl aliquot of eluate was used as template in a 20 µl total PCR reaction for 30 cycles. A 5 µl aliquote of each PCR reaction was resolved on a 2% precast agarose gel (Product No. P5722). The PCR marker (M) used (Product No. P9577) ranged from 50 bp to 2 kb.
Return to Top
Supplier Comparison of Preparation Time.
Supplier Comparison of Preparation Time
Figure 3. Comparison of preparation time. The prep time required using the GenElute Plant Genomic DNA Miniprep Kit compared to that of three kits from other suppliers.
The RNase treatment is not always required. The GenElute Plant Genomic DNA Miniprep Kit has been tested for use with corn, dianthus tissue culture, pepper, rice, soybean, tobacco, tomato and wheat. Other sources include Aspergillus parasiticus mold, potato tissue disks, snail and Arbuscular mycorrhizal fungal spores.
Order Information
Product Code Name Purifications per kit Technical Manual .pdf Short
Protocol.pdf
G2N10 GenElute Plant Genomic DNA Miniprep 10 X X
G2N70 GenElute Plant Genomic DNA Miniprep 70 X X
G2N350 GenElute Plant Genomic DNA Miniprep 350 X X
Return to Top