Life Science
Antibody Explorer
Automation
Cancer Research
Cell Culture
Cell Signaling and Neuroscience
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Nutrition Research
Obesity Research
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Your Favorite Gene - Search
Life Science Quarterly
PathFinder
 GenElute™ Plasmid Miniprep Kit
DNA and RNA Purification
Description: GenElute™ Plasmid Miniprep Kit offers a simple, rapid, and cost-effective method for isolating up to 20 µg plasmid DNA from E. coli cultures.
  • Fast – 24 preps in 30 minutes
  • Convenient – spin column format
  • Cost effective – more preps for the money
Procedure: Bacterial cells are harvested via centrifugation, subjected to a modified alkaline-SDS lysis procedure and the DNA adsorbed onto silica in the presence of high salts. Contaminants are then removed by a simple wash step. Bound DNA is eluted in water or Tris-EDTA buffer.
Application: Plasmid DNA is ready for immediate use in restriction enzyme digestion (Figure 1) cloning, PCR, transcription, conventional and automated sequencing (Figure 2).
GenElute Bacterial Genomic Flow Chart
Beneficial Features
Culture Volume
1 - 5 ml overnight bacterial culture
Yield
Up to 20 µg of plasmid DNA
Speed
30 minutes for 24 preps
Versatile
Contains additional wash buffer for use with EndA+ E. coli bacterial strains (e.g., HB101, JM101, BL21)
Specific
No detectable genomic DNA or RNA contamination
Safe
No phenol/chloroform required
Return to Top
Order Information
Product Code Culture Volume (ml) Plasmid
Yield (µg)
Time
Required
Number
of Preps
Technical Manual .pdf Short protocols .pdf
PLN10
PLN70
PLN350
1 to 5 30 to 20 Hands On: < 10 min.
Total: < 30 min.
10
70
350
X
X
X
X
X
X
PLD35
PLD140
5 to 40 25 to 200 Hands On: < 10 min.
Total: < 40 min.
35
140
X
X
X
X
PLX15
PLX50
25 to 130 200 to
1,200
Hands On: < 10 min.
Total: < 40 min.
15
50
X
X
X
X
Return to Top
Sample Data
Plasmid DNA purified with GenElute Plasmid Miniprep Kit is suitable for restriction enzyme digestions.
Figure 1. Restriction digestions of pUC19 purified from JM109 using the GenElute Plasmid Miniprep Kit. Lane 1 was an undigested plasmid, lanes 2-4 were cut with Bam HI, Eco RI and Pst I respectively. Lane M was 1 kb ladder (11 fragments from 0.5 to 10 kb). Electrophoresis was on a 1% agarose gel.
NAP Plasmid Gel
Return to Top
High-quality plasmid DNA for Automated Sequencing.
Figure 2. Electropherogram revealing >600 bases of sequence from pUC-TMV using GenElute Plasmid Miniprep Kit. Cycle Sequencing was performed using 500 ng of template, a T7 sequencing primer, and ABI Big DyeTM terminator chemistry. Sequencing reactions were resolved on an ABI Prism 377 XL instrument with a 48 cm gel cassette containing 4.5% AutoPAGE Plus acrylamide at 2.4 kV, 48°C for 7 hours.
Click Image to View Sequence in its Entirety
Return to Top