Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 GenElute™ mRNA and GenElute Direct mRNA Miniprep Kit
DNA and RNA Purification
Description: Procedures such as cDNA synthesis, expression profiling, and others, require separation of mRNA from the vastly more abundant rRNA and tRNA. The GenElute mRNA Kits provide convenient procedures for isolating polyadenylated mRNA from previously prepared total RNA, or directly from mammalian cells and tissues.
For direct mRNA preparation, cells or tissues are disrupted with SDS/proteinase K digestion to release RNA and eliminate RNases. Both kit types use oligo dT30 covalently linked to 1 µm polystyrene beads to capture polyadenylated mRNA by hybridization. The polystyrene beads remain suspended during hybridization, eliminating the need for mixing or rocking, as is common for cellulose or magnetic particles. Polystyrene was also chosen because oligo (dT) polystyrene beads yield cleaner mRNA with fewer stringent washing steps than does the more commonly used oligo (dT) cellulose (2 or 3 wash steps versus 10 or more). With the GenElute kits, mRNA-bead complexes are washed on a microcentrifuge spin filter, and eluted into 10 mM Tris-HCl, pH 7.5. mRNA prepared with either kit is suitable for a variety of downstream applications such as Northern Blotting (Figure 4), Expression Array or Chip Hybridizations, and cDNA Synthesis and Library Construction.
GenElute mRNA Miniprep Flow Chart
Protocol for GenElute
mRNA Miniprep Kits
Kit Contents
Features and Benefits
Fast
Prep time is 40 minutes for poly A+ mRNA from total RNA or 60 minutes directly from cells and tissues.
Convenient
mRNA captured on polystyrene beads in 10 minutes, with no mixing or rocking. Beads washed on microcentrifuge spin filters.
Yield
Higher than all competing products.
Return to Top
GenElute mRNA From Total RNA Kits
Product Code
Name
Purifications per Kit
Capacity
(µg Total RNA)
Technical Manual .pdf
Short Protocols .pdf
GenElute mRNA Miniprep Kit
10
5-500
GenElute mRNA Miniprep Kit
70
5-500
GenElute Direct mRNA Kits
Product Code
Name
Purifications per Kit
Capacity
Technical Manual .pdf
Short Protocols .pdf
GenElute Direct
mRNA Miniprep Kit
10
up to 107    up to 50mg
cells         tissues
GenElute Direct
mRNA Miniprep Kit
70
up to 107    up to 50mg
cells         tissues
Return to Top
Kit Contents
GenElute mRNA From Total RNA Kits
  • Water, Molecular Biology Grade
  • 2x Binding Solution
  • Oligo (dT) Polystyrene Beads
  • Wash Solution
  • Elution Solution
  • Spin Columns with Tubes
  • Collection Tubes
GenElute Direct mRNA Kits
    Lysis Solution
  • Proteinase K and Hydration Reagent
  • 5 M NaCl
  • Oligo (dT) Polystyrene Beads
  • Wash Solutions 1 & 2
  • Elution Solution
  • Filtration Columns with Tubes
  • Spin Columns with Tubes
  • Collection Tubes
Return to Top
Preparation Time Comparison
GenElute mRNA Prep Time
Figure 1. Comparison of the preparation time, to isolate mRNA from total RNA or direct from cells or tissues, using GenElute mRNA Miniprep Kit and other commercially available kits. Each kit was prepared according to the procedure supplied by the vendor.
Return to Top
RNA Yield
GenElute mRNA Yield
Figure 2. Total RNA was prepared from Hek293 cells by a silica-binding method, using TriReagent®. mRNA was subsequently prepared from total RNA preparations using commercially available kits, following the procedures supplied by each vendor. RNA yield was measured using the RiboGreenTM RNA Quantitation Kit (Molecular Probes).
Return to Top
Specific mRNA yield
GenElute Specific mRNA Yield
Figure 3. mRNA was prepared from total RNA as in Figure 2, and twenty percent of each preparation was analyzed by Northern blot hybridization with 32P-labeled probes. Hybridization to human HGPRT, human p53, and mouse p53 mRNAs were quantified with a Perkin Elmer Instant Imager.
Return to Top
mRNA is suitable for Northern Blots
GenElute mRNA Northern Blot
Figure 4. Northern blot comparison of mRNA prepared with GenElute mRNA kits from other suppliers. Duplicate mRNA samples were prepared from 5 x 106 Hek293 cells (top left panels) or 25-35 mg mouse liver (top right panels) with Sigma's GenElute Direct mRNA Miniprep Kit (S) or with another commercially available direct mRNA miniprep kit (A, D, I, P, & Q). An optional release and rebind procedure (described in the Manual) was included for preparation S+: RNA was hybrid-captured onto oligo (dT) beads, released into fresh lysis solution, and re-captured onto the same beads before washing and eluting mRNA. The release and rebinding steps were omitted for S- & Sx2 preparations. Sx2 preparations were repurified after the final elution with fresh oligo (dT) beads (as described in the Manual) . Equal portions of each mRNA preparation, equal to the amount from 1 x 106 cells or 10 mg liver, were evaluated by Northern blot hybridization with 32P-labeled RNA probes for p53, hypoxanthine-guanine phosphoribosyl transferase (HGPRT), c-myc, and/or glyceraldehyde 3-phosphate dehydrogenase (GAPDH) mRNA.
For the bottom panels, total RNA was first prepared from Hek293 cells and from mouse liver. Duplicate mRNA samples were then prepared from 100 µg of these total RNA preparations with Sigma's GenElute mRNA Miniprep Kit (S) or with another commercially available mRNA miniprep kit (Q, A, I, P, D). Samples Sx2 were repurified after elution. Twenty percent of each preparation was evaluated by Northern blot as above. In lanes T, 5 and 2 µg of the original total RNA from cells or 10 and 5 µg of total RNA from liver were analyzed for comparison.
Return to Top