Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 PhasePrep™ BAC DNA Kit
DNA and RNA Purification
Description: PhasePrep™ BAC DNA Kit offers a highly scalable, rapid, cost-effective method for isolating high molecular weight plasmids such as Bacterial Artificial Chromosomes (BACs) from recombinant E. coli cultures.
Procedure: The kit utilizes a phase separation procedure that can be scaled to micro, mini, midi, or maxi prep sizes. Sufficient reagents are provided for 300 micro, 180 mini, 30 midi, or 15 maxi preps.
Application: The purified BAC DNA contains very low levels of endotoxins and is ready for immediate use in Automated Sequencing (Figure 1), PCR, Restriction Enzyme Digestion (Figure 2), Cloning and other common applications.
GenElute PhasePrep BAC DNA Flow Chart
Beneficial Features
Starting Material
5, 40, 250 or 500 ml overnight culture for micro, mini, midi or maxipreps, respectively
Yield
Typical DNA yields of 2 - 100 µg from 5 - 500 ml of overnight cultures
Speed
Less than 1 hour; no tedious gravity-flow columns
Safe
No phenol/chloroform extraction required
Versatile
Allows possible micro to maxi preps with the same kit
Return to Top
Order Information
Product Code Name Culture Volume Expected Yield Technical Manual .pdf Short protocols .pdf
NA0100 PhasePrep BAC DNA Kit 5 - 500 ml 2 - 100 µg X X
Return to Top
Sample Data
High-quality BAC DNA for Automated Sequencing.
Figure 1. BAC DNA was purified with the PhasePrep BAC DNA Kit and sequenced with a custom primer using BigDye™ Terminator Chemistry. Sequencing reactions were resolved on an ABI 3700 sequencing instrument.
Data provided by the Genome Sequencing Center at Washington University, St. Louis
Click Image to View Sequence in its Entirety
Return to Top
Purified BAC DNA is suitable for restriction digestion.
Figure 2. Restriction digestion of BAC DNA isolated with the PhasePrep BAC DNA Kit and two other suppliers (C and Q). BAC DNA samples were purified from overnight cultures of E. coli HB101b transformed with a pbelloBAC11 clone. Approximately 1 g of DNA from each sample was digested with 20 units of Eco R V at 37°C for 4 hours. One half of each digestion (approximately 0.5 micrograms) was separated by overnight electrophoresis in a 1% agarose gel. Lanes 1 to 8 were BAC DNA purified with Sigma kit, lane 9 was BAC DNA purified with Supplier C's kit, and lane 10 was BAC DNA purified with Supplier Q's kit. The molecular marker used at the first and last lanes was a 1kb DNA Ladder (M) (Product Code D0428)  
Return to Top