Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Sequencing Clean-Up Quick Reference Guide
DNA and RNA Purification
Click on the product for more information
Product Speed Starting Material Product Description Technology Applications
UltraClear™ Sequencing Reaction Clean-Up Kit Flexible based on user interface ≤10 µl sequencing reaction Removes unincorporated dye terminators, including BigDye version 3.1, dNTPs, residual salts and other interfering components from sequencing reactions Ultrafiltration membrane purification in a centrifugation format Fluorescent Sequencing
SigmaSpin™
96-Well Post Reaction Clean-Up Plates
< 8 minutes 10 to 20 µl sequencing reaction Removes unincorporated dyes, including Big Dye 3, excess salts and other interfering components from sequencing reactions Resin based purification in a centrifugation format Fluorescent Sequencing
SigmaSpin™
96-Well Post Reaction Clean Up Columns
< 8 minutes 10 to 20 µl sequencing reaction Removes unincorporated dyes, including BigDye 3, excess salts and other interfering components from sequencing reactions Resin based purification in a centrifugation format Fluorescent Sequencing and Microarray Analysis