Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 SigmaSpinTM Post-Reaction Clean-Up Columns
Sequencing header
Description:
SigmaSpin Post-Reaction Clean-Up Columns SigmaSpin Sequencing Reaction Clean-Up Columns are ideal for lower throughput applications, such as clean up of probe labeling reactions or small numbers of sequencing reactions.
These columns can accept sample volumes up to 100 µl. Each column comes with a collection tube to collect the DNA during centrifugation.
SigmaSpin 96-Well Sequencing Reaction Clean-Up Plates
SigmaSpin 96-Well Sequencing Reaction Clean-Up plates provide a fast, simple, and highly efficient method for removing unincorporated dyes, excess salts, and other interfering reaction components. Each plate is packed with a size-exclusion resin, prehydrated with molecular biology grade water. Unique plate design incorporates drip directors to eliminate contamination between samples. SigmaSpin has been tested in high-throughput genome centers and core facilities with ABI Prism 3700, 3100, 377 and Amersham Biosciences MegaBACE automated DNA sequencers. SigmaSpin is compatible with all standard terminator chemistries including BigDye, DYEnamic ET and Thermo Sequenase II.
SigmaSpin Sequencing Reaction Clean-Up
SigmaSpin Plates and Columns
SigmaSpin 96-Well Clean-Up Plate
Features and Benefits:
Versatile
Compatible with all standard terminator chemistries and all commercially available DNA sequencers.
Fast
Quick and easy protocol requires 6 minutes total centrifugation.
High Performance
Pre-qualified size-exclusion resin guarantees optimum performance.
Recovery
Load and recover sample volumes down to 5 µl
Precision
Unique drip directors prevent cross-contamination between samples
SigmaSpin Removes Interfering Reaction Components and Eliminates Dye Blobs
SigmaSpin Seq Data
Figure 1. Sequencing reaction was purified using a SigmaSpin 96-well plate. pFLAGTM MAC plasmid (E 5644) was sequenced using BigDyeTM Terminator v 3.0 chemistry (20 µl total reaction volume; BigDye premix diluted 1:1 with SeqSaverTM [S3938]). Data was generated on an ABI Prism. 3700 DNA Analyzer with POP-6TM polymer and 10X CE Buffer (B4930). Phred 20 > 600.