Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 SpyLine™ Poly A+ Capture Kit
DNA and RNA Purification
The SpyLine™ Poly A+ Capture Kit offers a simple, rapid, and cost-effective method for isolating poly A+ mRNA from cultured mammalian cells for direct RT-PCR. This kit features the SpyLine Poly A+ Plate, which is a 96-well PCR plate coated with oligo(dT) for selective capture of poly A+ mRNA from crude mammalian cell lysates. RT-PCR reagents can be added directly to this capture plate for subsequent RT-PCR cycling and analysis, or the mRNA can be eluted from the plate if desired.
Lyse cells in culture plate
Transfer lysate to capture plate
Hybridize for 30-90 minutes
Wash
Perform RT-PCR directly in capture plate, or elute mRNA for other applications
Beneficial Features
Sensitive
Superior tool for knockdown measurement and gene expression analysis.
Flexible
Isolate mRNA from a wide variety of cell dilutions - even from a single cell.
Consistent
Extremely low well-to-well and plate-to-plate variability.
Automatable
96-well plate format suitable for use on high-throughput platforms.
Convenient
Capture and amplify mRNA in a single plate.
Simple
Rapid, easy-to-follow protocol.
Return to Top
Ordering Information
Product Code
Product Name
Available Sizes
Technical Manual .pdf
SpyLine Poly A+ Capture Kit
1 x 96-well plate
SpyLine Poly A+ Capture Kit
4 x 96-well plate
X
Superior sensitivity for siRNA knockdown
Figure 1. Target vs. dCT for SpyLine vs. Supplier Q. The figure above is a comparison of the change in Ct from the siRNA knockdown cells to the Non-Targeting siRNA cells. Plated HeLa cells were transfected with either siRNA transfection reagent mixture or non-targeting siRNA #1 reagent. After overnight incubation, the transfection media was removed, and the cells were lysed. One plate was purified by the SpyLine Poly A+ Capture Kit, while the other was purified using Supplier Q’s 96-well total RNA purification kit. Quantitative RT-PCR was run on the 2 sets of purified RNA using Applied Biosystems TaqMan® Gene Expression Assays.
Flexible enough for a variety of cell dilutions
Figure 2. Ct vs. cell number for decreasing amount of cells. A series of cell dilutions was prepared, beginning with 10,000 cells and decreasing by ten-fold increments down to 1 cell. SURF-4 mRNAs were then amplified by quantitative RT-PCR.
Figure 3. Quantitative RT-PCR of mRNA isolated from a series of decreasing cell dilutions. Cell dilutions were prepared in replicate, in decreasing 10-fold increments from 10000 cells to 1 cell. SURF-4 mRNAs were then amplified by quantitative RT-PCR. As depicted above, message can be clearly detected all of the way down to a single-cell input.
Consistent results - extremely low well to well and plate to plate variability
Figure 4. Well to well variability and plate to plate variability of the SpyLine Poly A+ Capture Kit. mRNA was isolated and captured utilizing the SpyLine standard protocol. Quantitative RT-PCR was set-up in the SpyLine Poly A+ Capture plate and was run targeting SURF-4 mRNA. Comparison of the two plates shows very little variability on each plate (standard deviation ~0.5 Cts), and very little variability from plate to plate (standard deviation ~0.5 Cts).
Back to mRNA Quick Reference Guide
Back to DNA and RNA Purification