Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 TRI® Reagent RNA Isolation Reagent
DNA and RNA Purification
Description: TRI Reagent is an improved version of the single-step total RNA isolation reagent developed by Chomczynski (1). The RNA isolation method based on this reagent is widely recognized and proven for RNA applications and is supported by a substantial publication list (2,3,4,5). It is ideal for quick, economical, and efficient isolation of total RNA or the simultaneous isolation of RNA, DNA and proteins from samples of human, animal, plant, yeast, bacterial and viral origin.
Beneficial Features
Yield
Better yields than traditional guanidine thiocyanate or cesium chloride methods
Versatile
Works with many sources including human, plant, yeast, bacterial or viral
Convenience
Three convenient formulations
Efficient
Easily scalable RNA isolation
Return to Top
Tri Reagent RNA Isolation Reagent
Product Number
Product Description
Product Size
Tri Reagent RNA, DNA and Protein Isolation Reagent
25, 100 and 200 ml
Tri Reagent BD
25, 100 and 200 ml
Tri Reagent LS
25, 100 and 200 ml
1-Bromo-3-Chloropropane (BCP)
200 ml
Tri Reagent Formulations
Product Name
Sample Type
Sample Volume
TRI Reagent Volume
TRI Reagent
Tissues, cultured adherent cells, cell pellets
up to 100 mg tissue, 107 cells or 102 cm plate area
1 ml
TRI Reagent BD
Whole blood, plasma, serum
0.25 ml blood derivatives
0.75 ml
TRI Reagent LS
Cell suspension, CSF, amniotic fluid
0.25 ml fluid samples
0.75 ml
Typical RNA Yield
Tissue
Yield
(µg RNA/mg tissue)
Cultured Cells
Total RNA Yield
(µg RNA/mg tissue)
Liver
6 - 10
Epithelial
8 - 15
Spleen
6 - 10
Fibroblast
5 - 7
Kidney
3 - 4
Skeletal Muscle
1 - 1.5
Brain
1 - 1.5
Placenta
1 - 4
Total RNA was prepared from HeLa cells using TRI Reagent from Sigma and other various suppliers
Figure 1. Total RNA from HeLa cells was prepared using Tri Reagents from Supplier R (Lane 1), Sigma (Lane 2) and Supplier L (Lane 3). A 2 µl aliquot out of 200 µl total RNA was analyzed on a 1% agarose gel. RNA Marker (M) used ranged from 0.2 bp - 10 kb (Product Code R7020). TRI Reagent RNA Isolation Reagent
References
1. Chomczynski, P. and Sacchi, N., Single-step method of RNA isolation by acid guanidinium thiocyanate-phenol-chloroform extraction. Anal. Biochem. 162: 156 (1987).
2. Chomczynski, P., A reagent for the single-step simultaneous isolation of RNA, DNA and proteins from cell and tissue samples. BioTechniques 15: 532-537 (1993).
3. Mackey, K. and Chomczynski, P., Long-term stability of RNA isolation reagents. J. NIH Res. 8: 72 (1996).
4. Wilfinger, W., et al., Effect of pH and ionic strength on the spectrophotometric assessment of nucleic acid purity.BioTechniques 22: 474-481(1997).
5. Chomczynski, P. and Mackey, K., Modification of the Tri Reagent. procedure for isolation of RNA from polysaccharide- and proteoglycan-rich sources.BioTechniques 19: 924-945(1995).