Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 UltraClear™ Sequencing Reaction Clean-Up Kit
DNA and RNA Purification
The Advantage is Clear with Sigma’s
UltraClear™ Sequencing Reaction Clean-Up Kit
The UltraClear Sequencing Reaction Clean-Up Kit offers a rapid and simple method for the clean-up of DNA sequencing reactions in a 96-well format. The UltraClear plate uses ultrafiltration membranes to separate low molecular weight contaminants,such as unincorporated dye terminators, dNTPs, and residual salts from the sequencing reaction products.
UltraClear Protocol
Dilute Sequencing Reactions
Load Ultra Clear Plate
Centrifuge
Add 40 µl Sequencing Solution
Resuspend the Purified Sequencing Products
Transfer Sequencing Product to an Appropriate Injection Plate for each configuration.
Features and Benefits
  • Excellent quality – PHRED q>20 scores greater than 850 bases (See Data)
  • Scientifically engineered to use less BigDye® version 3.1 chemistry and save money
  • Optimized centrifugation procedures for maximum flexibility
  • Partial 96-well plate can be used and stored at room temperature
Order Information
Product Code Product Name Size
UC9601 UltraClear Sequencing Reaction Clean-Up 1 x 96
UC9604 UltraClear Sequencing Reaction Clean-Up 4 x 96
back to top
High Quality Sequence Data with UltraClear Kit
back to top
Excellent PHRED q>20 Scores Compared to Competition
Data quality analysis based on PHRED q>20 scores. Samples analyzed on the ABI Prism® 3700 under standard run conditions. M13 template was prepared using 1/8 BigDye Terminator v3.1.
back to top
Optimized Protocol Accommodates a Variety of Spin Forces and Times
Spin Force Time Avg. PHRED 20 SD
1,000 x g 30 minutes 887 11
1,500 x g 20 minutes 870 15
2,500 x g 15 minutes 880 16
3,500 x g 10 minutes 859 39
Average PHRED q >20 scores for each spin force/time configuration tested. N=8
back to top
back to DNA and RNA Purification