Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Plasmid Purification Kits
Genomic DNA
Extract-N-Amp
Plasmid Partner
Post Reaction Cleanup
Sequencing Clean-Up
Total RNA
mRNA
RNA Reagents
Product Guide
Products by Application
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 mRNA Quick Reference Guide
DNA and RNA Purification
Click on the product for more information
Product Yield Speed Starting Material Product Description Technology Applications
SpyLine™ Poly A+ Capture Kit Captured mRNA for direct RT-PCR on plate (optional elution if desired) 40-90 minutes 1000-50000 cells per well Isolation of mRNA from cultured mammalian cells for direct RT-PCR Capture of poly A+ mRNA onto 96-well PCR plate coated with oligo dT; optimized for high-throughput isolation RT-PCR (in plate), northern blots and microarray analysis (after optional elution)
GenElute™ Direct mRNA Miniprep Kit Up to 5 µg 60 minutes Up to 107 mammalian cells or
50 mg of tissue per prep
Isolation of mRNA from mammalian cells and tissues Hybrid-capture onto oligo dT-polystyrene beads; wash & elute in a spin column format RT-PCR, northern blots and microarray analysis
GenElute™ mRNA Miniprep Kit 1-5% of starting total RNA 40 minutes 5-500 µg total RNA Isolation of mRNA from total RNA Hybrid-capture onto oligo dT-polystyrene beads; wash & elute in a spin column format RT-PCR, northern blots and microarray analysis