Automation
Biological Buffers
Cancer Research
Cell Culture
Cell Signaling and Neuroscience
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 QuantiGene® 2.0 RNA Quantification System Process & Protocol Diagram
Gene Expression & Analysis
Sigma-Aldrich now offers the QuantiGene® RNA Quantification System to measure gene expression knockdowns. As a complement to the MISSION™ shRNA Library, the QuantiGene RNA Quantification System and probe sets provide researchers with the tools to accurately validate RNAi gene knockdown assays. The probe sets and reagent kit are necessary to use the QuantiGene system in validating gene expression.
The QuantiGene® RNA Quantification System is a sandwich nucleic acid hybridization assay that provides a unique approach for RNA detection and quantification by amplifying the reporter signal using branched DNA (bDNA) technology. The bDNA technology allows quantification of target-specific RNA molecules directly from a variety of samples types without RNA purification or target amplification. By measuring RNA directly from crude cell lysates or tissue homogenates, the assay avoids variations or errors inherent to extraction and amplification of target sequences.
Process Diagram
Step 1
Lyse cells to release mRNA in the presence of target probes. During overnight incubation, probe set anchors the specific target sequence to the capture plate through multiple CEs binding to the capture plate.
QuantiGene® 2.0 RNA Quantification System 
Process
Step 2
Pre-amplifier and amplifier probes (not shown in image) are added in separate steps for hybridizing to the LEs. Incubation allows the label probe to hybridize to the bDNA.
Step 3
Addition of chemiluminescence substrate yields a luminescent signal that is proportional to the amount of mRNA present in the sample.
QuantiGene® is a registered trademark of Bayer.
back to QuantiGene
back to Gene Expression & Analysis