Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
Product Highlights
Product Lines
Key Resources
PCR
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Product Lines
Nucleic Acid Electrophoresis
Agarose and Acrylamide | Dyes, Loading Buffers, and Nucleic Acid Gel Stains | Markers and Ladders | Running Buffers | Equipment and Accessories
Agarose and Acrylamide
  Agarose
Pre-cast agarose gels
Acrylamide
Agarose
Sigma-Aldrich offers a range of molecular biology grade agarose products from routine use to wide range and low melt point agarose. Each lot of products is rigorously tested to ensure reproducibility from one lot to the next. Use the following table to select the best product for your application.
Product # Product Name Recommended DNA size range Specific applications Gelling Temp
(°C)
Melting Temp
(°C)
EEO Moisture Gel Strength (g/cm2) Sulfate
A9539 Routine Use 0.5-25 kb analysis, purification 36 + 1.5   0.09-0.13 <10% >1200 1% gel <0.15%
A4718 High Resolution 200-800 bp small NA fragments <35 <75 <0.05 <10% >300 3% gel  
A2929 Pulsed-field gel electrophoresis 10-2000 kb field inversion electrophoresis 42+ 1.5   0.04-0.07 <8% >2400 1.5% gel <0.10%
A3054 Pulsed-field sample prep N/A gel plugs, lysis and digestion of embedded DNA 26-30 <70 <0.10 <10% >400 1% gel <0.15%
A9414 Low melting 0.5-25 kb cloning and isolation from agarose 26-30 <65 <0.10 <10% >200 1% gel <0.10%
A2790 Wide range 50-1000 bp small NA fragments <35 <65 <0.15 <10% >500 4% gel <0.15%
A7431 Wind range: standard (3:1) 50-1000 bp small NA fragments; strong flexible gels 32.5-38 <90 <0.13 <10% >1400 4% gel <0.15%
back to top
Pre-cast agarose gels
Pre-cast agarose gels are designed for high-resolution separation of nucleic acids, producing sharp DNA and RNA bands with low background fluorescence. Pre-cast gels are individually foil-sealed in plastic trays and can be electrophoresed right in the plastic tray. They provide the ultimate in convenience for nucleic acid agarose electrophoresis.
Product Name Technical Bulletin Product #
1.0% Agarose in TBE buffer with 0.5 µg/ml Ethidium bromide 8-well (portrait orientation) P5472
1.0% Agarose in TBE buffer with 0.5 µg/ml Ethidium bromide 20-well (landscape orientation) P5722
1.0% Agarose in TBE buffer with 0.5 µg/ml Ethidium bromide 24-well (2x12-well portrait orientation) P5972
4.0% Agarose in TBE buffer with 0.5 µg/ml Ethidium bromide 8-well (portrait orientation) P5597
4.0% Agarose in TBE buffer with 0.5 µg/ml Ethidium bromide 20-well (landscape orientation) P5847
4.0% Agarose in TBE buffer with 0.5 µg/ml Ethidium bromide 24-well (2x12-well portrait orientation) P6097
1.25% Agarose in MOPS for RNA electrophoresis 8-well (portrait orientation) P6222
back to top
Acrylamide
Acrylamide is used for both protein and nucleic acid electrophoresis. The following table can be used to determine the appropriate acrylamide:bis acrylamide concentration for your application.
Product Name Technical Bulletin Product #
Acrylamide, for molecular biology, minimum 99% A9099
Acrylamide, for mutation detection A5934
Gel Concentration (% w/v) Effective range of separation (bp) Xylene Cyanol (bp) Bromophenol Blue (bp)
3.5 1000-2000 460 100
5.0 50-500 260 65
8.0 60-400 160 45
12.0 40-200 70 20
15.0 25-150 60 15
20.0 6-100 45 12
Acrylamide:bis-acrylamide ratio 29.1, 1X TBE buffer
Size of the double stranded DNA, which comigrates with the dye.
back to top