PathFinder
 Xanthohumol
Nutrition Research
Xanthohumol
from Hop (humulus lupulus)
Xanthohumol is a prenylated chalcone found naturally in Hop (humulus lupulus). This prenylated flavonoid has been shown to be a potent bioactive compound. Xanthohumol has been shown to have antiproliferative and cytotoxic effects in human cancer cell lines.1,2 It has also been displayed to inhibit diacylglycerol acetyltransferase (DGAT)3 and human P450 enzymes. Xanthohumol inhibits the expression of HIF-1α and VEGF under hypoxic conditions.
Higher antioxidant activity is reported for prenylchalcones than for prenylflavanones in the Cu2+-mediated oxidation of LDL, suggesting a relation between structure and function.4 Also, many chalcones suppress tumor promotion more effectively than flavonoids themselves.5
Quantities of xanthohumol found in Hop are too small to have any biological effects under normal consumption amounts.
Product Number: X0379
Line drawing of Xanthohumol chemical structure
Molecular Formula: C21H22O5
Molecular Weight: 354.40
Preparation Instructions
Soluble in DMSO at 25 mg/ml. Also soluble in ethanol at 10 mg/ml.
Storage Conditions
Store at 2-8 °C. Under these conditions the product is stable for at least 2 years.
References
  1. Miranda, C.L. et. al., Antiproliferative and cytotoxic effects of prenylated flavonoids from hops (Humulus lupulus) in human cancer cell lines. Food Chem. Toxicol. 37, 271-285, (1999)
  2. Delmulle, L. et. al., Antiproliferative properties of prenylated flavonoids from hops (Humulus lupulus L.) in human prostate cancer cell lines. Phytomedicine 13, 732-734, (2006)
  3. Tabata, N. et. al., Xanthohumols, diacylglycerol acyltransferase inhibitors, from Humulus lupulus. Phytochemistry 46, 683-687, (1997)
  4. Miranda, C.L., et. al., Antioxidant and prooxidant actions of prenylated and nonprenylated chalcones and flavanones in vitro. J. Agric. Food Chem 48, 3876-3884, (2000)
  5. Dimmock, J. R., et. al., Bioactivities of chalcones. Curr. Med. Chem. 6, 1125-1149, (1999)