Product Lines
Cytokines
Gene Regulation
Lipid Signaling
Neurotransmission
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Gene Regulation and Obesity Research
Obesity Research
Gene regulation, specifically regulation of the leptin receptor and JAK/STAT signal transduction pathway can also influence obesity. Leptin is the product of the obese (ob) gene and is produced predominantly in white adipose tissue. The leptin receptor (Ob-R) isolated by expression cloning, is encoded by the diabetes (db) gene. The arcuate nucleus (ARC) of the hypothalamus serves as the leptin signaling center. Leptin receptor activation stimulates STAT1, STAT3 and STAT5 tyrosine phosphorylation in vitro and STAT3 tyrosine phosphorylation in vivo. The Ob-Rb, long leptin receptor isoform, acts as a potent JAK/STAT signaling activator. Ciliary neurotrophic factor (CNTF) appears to act similar to leptin. CNTF administration has been shown to reduce adiposity much like leptin. Systemic administration of CNTF activates the JAK/STAT pathway in the ARC. Unfortunately, Axokine an analog of CNTF, showed disappointing results in clinical trials for the treatment of obesity. However, there is still promise for gene regulation therapies in the treatment of obesity.
Control Peptides for Transcription Regulators
Product #  Product Name Notes Add to Cart
S4315 STAT1α trifluoroacetate salt Control peptide for the STAT1α rabbit polyclonal antibody.
S5065 [pSer727]-STAT1, mouse trifluoroacetate salt Positive control peptide for the [pSer727]-STAT1 rabbit polyclonal antibody.
S5190 [Ser727]-STAT1 trifluoroacetate salt Negative control peptide for the [pSer727]-STAT1 rabbit polyclonal antibody.
S4065 [pTyr701]-STAT1 trifluoroacetate salt Positive control peptide for the [pTyr701]-STAT1 rabbit polyclonal antibody.
S4190 [Tyr701]-STAT1 trifluoroacetate salt Negative control peptide for the [pTyr701]-STAT1 rabbit polyclonal antibody.
S4440 STAT2 (830-849) trifluoroacetate salt Control peptide for the STAT2 rabbit polyclonal antibody.
S4565 STAT3 (733-748) trifluoroacetate salt Control peptide for the STAT3 rabbit polyclonal antibody.
S4815 [pTyr705]-STAT3, mouse trifluoroacetate salt Positive control peptide for the [pTyr705]-STAT3 (mouse) rabbit polyclonal antibody.
S4940 [Tyr705]-STAT3, mouse trifluoroacetate salt Negative control peptide for the [pTyr705]-STAT3 (mouse) rabbit polyclonal antibody.
S5315 [pSer727]-STAT3 trifluoroacetate salt Positive control peptide for the [pSer727]-STAT3 rabbit polyclonal antibody.
S4690 STAT4 trifluoroacetate salt Positive control peptide for the STAT4 rabbit polyclonal antibody.
Antibodies to Transcription Regulators
Product #  Product Name Species Reactivity Immunogen Application Add to Cart
S2190 Anti-STAT1α, Rabbit affinity isolated antibody Mouse Mouse STAT1α Immunoblotting
S4558 Anti-phospho-STAT1 (pSer727), Rabbit IgG fraction of antiserum Human, mouse, rat Synthetic phosphopeptide corresponding to amino acids 718-730 of human STAT1 that contains serine 727 Indirect Immunoblotting
S2315 Anti-phospho-STAT1 (pTyr701), Rabbit affinity isolated antibody Human, mouse Synthetic phosphopeptide derived from the region of human STAT1 that contains tyrosine 701 Immunoblotting
S2565 Anti-phospho-STAT1 (pTyr701), mouse, (Anti-mSTAT 1 (pT701), Rabbit affinity isolated antibody Mouse, human (weaker), rat ISynthetic phosphopeptide derived from the region of murine STAT1 that contains tyrosine 701 Immunoblotting
S4183 Monoclonal Anti-phospho-STAT3 (pTyr705), Clone 9E12; Mouse purified immunoglobulin Human, mouse Synthetic phosphopeptide containing the AP[pY]LK motif of human STAT3 Indirect Immunoblotting, Immunoprecipitation
S2690 Anti-phospho-STAT3 (pTyr705), Rabbit affinity isolated antibody Human, mouse, rat Synthetic phosphopeptide derived from the region of human STAT3 that contains tyrosine 705 Immunoblotting
S5683 Anti-STAT1, Rabbit IgG fraction of antiserum Human, mouse GST fusion protein containing amino acids 714-750 of the C-terminal region of human STAT1, expressed in E. coli Indirect Immunoblotting, Immunoprecipitation
S5808 Anti-STAT2, Rabbit IgG fraction of antiserum Human, mouse GST fusion protein containing amino acids 671-806 of the C-terminal region of human STAT2, expressed in E. coli Indirect Immunoblotting, Immunoprecipitation
S5933 Anti-STAT3, Rabbit IgG fraction of antiserum Human, mouse, rat GST fusion protein corresponding to amino acids 688-722 of human STAT3, expressed in bacteria Indirect Immunoblotting, Immunoprecipitation
S5558 Anti-STAT4, Rabbit affinity isolated antibody Mouse, human, rat Synthetic peptide corresponding to amino acids 732-749 of mouse STAT4 Indirect Immunoblotting
S6058 Anti-STAT5A, Rabbit IgG fraction of antiserum Human, mouse Synthetic peptide corresponding to amino acids 778-794 of human STAT5A Indirect Immunoblotting, Immunoprecipitation
S6183 Anti-STAT5B, Rabbit IgG fraction of antiserum Human, mouse Synthetic peptide corresponding to amino acids 774-787 of human STAT5B Indirect Immunoblotting, Immunoprecipitation
S4308 Monoclonal Anti-phospho-STAT5A/B, (pTyr694,699), Clone 8-5-2; Isotype IgG1, Mouse purified immunoglobulin Human, mouse, rat, bovine, sheep Synthetic phosphopeptide corresponding to amino acids 689-700 of human STAT5A and amino acids 694-705 of human STAT5B Indirect immunoblotting
S5058 Anti-phospho-STAT5A/B (pTyr694,699), Rabbit IgG fraction of antiserum Human, mouse, rat Synthetic phosphopeptide, corresponding to amino acids 687-698 of human STAT5 Immunoblotting
S6308 Anti-STAT6 (689-711), Rabbit IgG fraction of antiserum Human Synthetic peptide corresponding to amino acids 689-711 of human STAT6 Indirect Immunoblotting, Immunoprecipitation
S6433 Anti-STAT6 (787-804), Rabbit whole antiserum Human, mouse (weakly) Synthetic peptide corresponding to amino acids 787-804 of human STAT6 Immunoblotting
S5061 Anti-phospho-STAT6 (pTyr641), Rabbit affinity isolated antibody Human, mouse Synthetic phosphopeptide derived from the region of STAT6 that contains tyrosine 641 Immunoblotting
back to top
back to Obesity Research