PathFinder Cell Signaling Pathway
www.sigmaaldrich.com/pathfinder
Sigma-Aldrich
Launch PathFinder
to see a detailed, interactive version of this map
NOTE: PathFinder requires the Flash plug-in for proper viewing. It will launch in a separate pop-up window, please turn off any pop-up blockers for this web site.
Activation of PKA through GPCR
Copyright ©2006 ProteinLounge.com
 Activation of PKA through GPCR
Protein kinase A (PKA) is a family of cAMP-dependent protein kinase isozymes that phosphorylate cellular proteins including: Rap-1 (involved with regulation of the Ras/Raf/MEK/ERK pathway at the level of MEK), CREB (cAMP Response Element-Binding protein), RelA (p65) (a subunit of NFkappaB heterodimers), and L-type calcium channels. CAMP (cyclic-AMP) is an important second messenger that is generated from ATP by the actions of adenylate (adenylyl) cyclases (AC). Cells can regulate levels of cAMP induced by external signals through the activation of specific G-protein coupled receptors (GPCRs). GPCRs are receptors linked to heterotrimeric G-proteins composed of (1:1:1) ratios of isoforms of alpha, beta and gamma subunits. The subunits of G-proteins interact with and regulate the activities of various effector molecules such as adenylate cyclase(s). Specificity for activation of adenylate cyclase(s) begins with the specific GPCR receptor. The signal is further refined by the specific isoform composition of the receptor associated trimeric G-protein and the specific iso- form or type of the targeted adenylate cyclase. A variety of AC isozymes exist. Finally kinase anchoring proteins (AKAPs) target PKA to specific substrates and distinct subcellular compartments providing spatial and temporal specificity.
Adenylate (adenylyl) cyclase(s) are regulated, in part, by the balance of their exposure to specific isoforms of heterotrimeric G-protein alpha subunits; alphaGs (Gα-s) is stimulatory and alphaGi (Gα-i) is inhibitory. AC activity is also regulated by the action of phosphodiesterases that hydrolyze cAMP to AMP and phosphatases that dephosphorylated the phosphorylated PKA substrates.
References:
  1. Lerosey I., et. al. (1991) The cAMP-dependent protein kinase phosphorylates the rap1 protein in vitro as well as in intact fibroblasts, but not the closely related rap2 protein. Brioche. Biophys. Res Commun. 175, 430-436.
  2. Takahashi, N., et. al. (2002) Inhibition of the NF-kB transcriptional activity by protein kinase A. Eur. J. Biochem. 269, 4559-4565.
  3. Tasken K, and Aandahl E.M. (2004) Localized effects of cAMP mediated by distinct routes of protein kinase A. Physiol Rev. 84, 137-167.
  4. Zhong H., et. al. (1998) Phosphorylation of NF-kappa B p65 by PKA stimulates transcriptional activity by promoting a novel bivalent interaction with the coactivator CBP/p300. Mol Cell. 5, 661-671.
Back to top
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net