PathFinder Cell Signaling Pathway
www.sigmaaldrich.com/pathfinder
Sigma-Aldrich
Launch PathFinder
to see a detailed, interactive version of this map
NOTE: PathFinder requires the Flash plug-in for proper viewing. It will launch in a separate pop-up window, please turn off any pop-up blockers for this web site.
Beta-Adrenergic Signaling Pathway
Copyright ©2006 ProteinLounge.com
 Beta-Adrenergic Signaling Pathway
The catecholamines epinephrine (adrenaline) and norepinephrine (noradrenaline) are agonists for a family of G-protein coupled receptors (GPCRs) known as adrenergic receptors. There are three subfamilies of adrenergic receptors: alpha1 (a1), alpha2 (a2) and beta (b). The beta subfamily contains three subtypes: b1 (b1AR), b2 (b2AR), and b3 (b3AR) located predominantly in heart, lung and adipose tissue respectively.
Adrenergic receptors are GPCRs that link to trimeric G-protein. G-proteins are typically composed of alpha, beta and gamma subunits. Each subunit exists in multiple isotypes with differential specificity for effector signaling. Specificity of response to catecholamines is mediated in part by the Galpha subunits, but the Gbetagamma dimers are also involved in regulation. Beta adrenergic receptors favor interaction with heterotrimeric G-proteins that contain the Galphas (Gs) and Galphai (Gi) subunit. The Galphas subunits activate various isoforms of adenylate (adenylyl) cyclases. Consequently b-adrenergic receptors typically elevate the level of cyclic AMP (cAMP) an important mediator of cell signaling. Both Galphas and Galphai have been linked to the stimulation of Src family tyrosine kinases. Galphas and Gbetagamma (dimeric) subunits regulate L-type Ca2+ channels. Beta adrenergic receptors also interact with other G-protein Galpha isotypes to provide additional specificity to agonist binding within cell context. Beta adrenergic receptors of different isotypes use combinations of G-proteins subunits to integrate specific cellular pathways and responses.
References:
  1. Ma Y.C., and Huang X.Y., (2002) Novel signaling pathway through the beta-adrenergic receptor. Trends Cardiovasc Med. 12, 46-49.
  2. Ma Y.C., et. al. (2000) Src tyrosine kinase is a novel direct effector of G proteins. Cell. 102, 635-646.
  3. Huang J., et. al. (2004) Distinct roles for Src tyrosine kinase in beta2-adrenergic receptor signaling to MAPK and in receptor internalization. J. Biol. Chem. 279, 21637-42162.
  4. Zhong J., et. al. (2001) beta-Adrenergic receptor stimulation of L-type Ca2+ channels in rabbit portal vein myocytes involves both alphas and betagamma G protein subunits. J. Physiol. 531, 105-115.
Back to top
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net