PathFinder Cell Signaling Pathway
www.sigmaaldrich.com/pathfinder
Sigma-Aldrich
Launch PathFinder
to see a detailed,interactive version of this map
NOTE: PathFinder requires the Flash plug-in for proper viewing. It will launch in a separate pop-up window, please turn off any pop-up blockers for this web site.
ERK Signaling Map
Copyright ©2006 ProteinLounge.com
 ERK Signaling
Extracellular signal-regulated kinases 1 and 2 (ERK1 and ERK2) are often referred to together as ERK1/2. ERK1/2 are pleiotrophic mitogen activated serine/threonine protein (MAP) kinases with over 160 known substrates, found throughout the cell, that include other protein kinases, membrane receptors, cytoskeletal proteins, downstream effector kinases and transcription factors. The ERK1/2 substrates share the consensus motif X-Pro-Xaan-Ser/Thr-Pro-X. ERK1/2 regulates proliferation, differentiation, cell cycle processes, and survival, as well as many other cell processes. Excessive ERK1/2 activation induces apoptosis. Many of the oncogenic effects of Ras-GTPases are mediated through ERK1/2 activation.
The central ERK1/2 activation pathway initiates with the activation of MAP3 kinases such as Raf-1 and/or B-Raf (BRAF) by specific Ras-GTPases that are activated by specific GEFs such as EPAC and SOS. EPACs are activated by cAMP and SOS(s) are down-stream effectors of receptor tyrosine kinase (RTK) activation. Rap1-GTPase activates B-Raf and inhibits Raf-1 activation by Ras-GTPase. Activation of Raf-1 and/or B-Raf leads to the activation of the MAP2 kinases, MEK1 and MEK2 (MEK1/2), which in turn activate ERK1/2. ERK1/2 differentially phosphorylates several members of the 90-kDa ribosomal S6 kinase (pp90rsk) (RSK) family including RSK1, RSK2, MNK1/2 and MSK1/2. Together ERK1/2 and the RSKs activate a plethora of transcription factors including but not limited to CREB, c-Myc, c-Jun, c-Fos, and STAT.
References:
  1. Chuderland, D. and Seger, R. (2005) Protein-protein interactions in the regulation of the extracellular signal-regulated kinase. Mol. Biotechnol. 29, 57-74.
  2. Gonzalez, F. A. et. al. (1991) Identification of substrate recognition determinants for human ERK1 and ERK2 protein kinases. J. Biol. Chem. 266, 22159-22163.
  3. Nguyen, T. T. et. al. (1993) Co-regulation of the mitogen-activated protein kinase, extracellular signal-regulated kinase 1, and the 90-kDa ribosomal S6 kinase in PC12 cells. Distinct effects of the neurotrophic factor, nerve growth factor, and the mitogenic factor, epidermal growth factor. J. Biol. Chem. 268, 9803-9810.
  4. Robbins, D. J. et. al. (1994) MAP kinases ERK1 and ERK2: pleiotropic enzymes in a ubiquitous signaling network. Adv. Cancer Res. 63, 93-116.
  5. Yoon, S. and Seger, R. (2006) The extracellular signal-regulated kinase: multiple substrates regulate diverse cellular functions. Growth Factors. 24, 21-44.
Back to top
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net