to see a detailed, interactive version of this map
|
NOTE: PathFinder requires the Flash plug-in for proper viewing. It will launch in a separate pop-up window, please turn off any pop-up blockers for this web site.
|
Copyright ©2006 ProteinLounge.com
| |
Mismatch Repair Pathways in Eukaryotes
DNA mismatch repair (MMR) is a process that maintains DNA homeostasis by facilitating post-replication repair, policing homologous recombination events and protecting against genetic exchange between species. Loss of MMR function results in the accumulation of potential mutations, a mutator phenotype, and genetic instability. As a post-synthesis mismatch repair process, MMR increases the fidelity of DNA replication about 1000-fold. It functions when DNA polymerase exonuclease 3’→5’ proof-reading fails to repair base-pair mismatches. MMR-independent pathways that process some types of mismatches in DNA include nucleotide-excision repair (NER), base excision repair (BER) glycosylases, and the flap endonuclease FEN-1.
Eucaryotic DNA mismatch repair (MMR) is initiated when either a mutSalpha (mutSα) dimer, MSH2(MutS homolog 2):MSH3(MutS homolog 3), or a MutSbeta (mutSβ) dimer, MSH2:MSH6(MutS homolog 6), recognizes and binds to mismatched DNA. The MSH2:MSH6 dimer recognizes base-base mismatches, while both mutS dimers can recognize small loop insertions. Binding of these dimers to mismatch DNA triggers ATP-dependent interaction of another heterodimer with the MSH-mismatch DNA complex. The second interacting heterodimer is a mutL homolog composed of either a mutLalpha (mutLα), MLH1(mutL homolog 1):Pms1(post-meiotic segregation increased 1) or a mutLbeta (mutLβ), MLH1:MLH3(mutL homolog 3) heterodimer. The formation of a ternary mutS:mutL:mismatch DNA complex leads to strand discrimination and error removal by an undefined process. MMS involves other factors such as proliferating cell nuclear antigen (PCNA), replication factor C (RF-C), exonuclease 1 (EXO1), and DNA polymerases δ and ε. PCNA may facilitate the search for mismatched DNA by MSH2:MSH6. RF-C, which contains a DNA-dependent ATPase, is reported to load PCNA onto DNA.
|
References:
Antony, E. et. a. (2006) Contribution of Msh2 and Msh6 subunits to the asymmetric ATPase and DNA mismatch binding activities of Saccharomyces cerevisiae Msh2-Msh6 mismatch repair protein. DNA Repair (Amst). 5, 153-162.
Hsieh, P. (2001) Molecular mechanisms of DNA mismatch repair. Mutat. Res. 486, 71-87.
Jascur, T. and Boland, C.R. (2006) Structure and function of the components of the human DNA mismatch repair system. Int. J. Cancer. 119, 2030-2035.
Jun, S.H. et. al. (2006) DNA mismatch repair system. Classical and fresh roles. FEBS J. 273, 1609-1619.
Kunkel, T.A. and Erie, D.A. (2005) DNA mismatch repair. Annu. Rev. Biochem. 74, 681-710.
|
Back to top |
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net
|