PathFinder Cell Signaling Pathway
www.sigmaaldrich.com/pathfinder
Sigma-Aldrich
Launch PathFinder
to see a detailed, interactive version of this map
NOTE: PathFinder requires the Flash plug-in for proper viewing. It will launch in a separate pop-up window, please turn off any pop-up blockers for this web site.
Nuclear Receptor Activation by Vitamin A
Copyright ©2006 ProteinLounge.com
 Nuclear Receptor Activation by Vitamin A
All-trans-retinoic acid (RA), vitamin A, is a pleiotropic factor involved in a wide variety of vertebrate cell processes including embryonic development, morphogenesis, survival, cell growth and differentiation, and tissue homeostasis. The importance of vitamin A is demonstrated by the fact that it is teratogenic when present in excess or deficient levels. Retinol (all-trans-retinol) is converted to retinal (all-trans-retinal) by retinol dehydrogenases (EC 1.1.1.105). All-trans retinal is converted into all-trans-retinoic acid (RA) by retinal dehydrogenases (EC 1.2.1.36).
All-trans-RA is the primary retinoid that affects cell processes at the level of transcription. All-trans-RA is the physiological ligand for a family of nuclear transcription factors, retinoic acid receptors (RAR) that form functional dimers with a second family of nuclear transcription factors, the retinoid X receptors (RXR). RAR and RXR belong to the steroid/thyroid/retinoid nuclear receptor family. RAR is the dominant partner in the RAR:RXR heterodimer. The role of RXR is to synergistically enhance the transcription regulatory effect of RAR. The physiological ligand for the RXR transcription factors is thought to be 9-cis-retinoic acid, but this is not definitively established. RAR:RXR heterodimers bind to DNA via cognate recognition sequences and mediate or modify gene expression through complex context and cell-specific interactions with transcription machinery.
There are three known RAR isotypes, RARα, RARβ, and RARγ that have been found as a variety of isoforms, RARα1, RARα2, RARβ1/β3, RARβ2/β4, RARγ1, RARγ2. These isotypes result from combinations of gene splicing and alternative promoter initiation events. RXR also exists as three isotypes, RXRα, RXRβ, and RXRγ and various isoforms. The effect of all-trans-RA on a cell’s transcription activity is determined by the specific isotype/isoform combination of RAR and RXR. Together these factors provide an opportunity for a combinatorial number of cell, context and age specific responses to all-trans-RA.
References:
    Chambon, P. (2005) The nuclear receptor superfamily: a personal retrospect on the first two decades. Mol. Endocrinol. 19, 1418-1428.
    Lefebvre, P. et. al. (2005) Transcriptional activities of retinoic acid receptors. Vitam. Horm. 70, 199-264.
    Mark, M. et. al. (2006) Function of retinoid nuclear receptors: lessons from genetic and pharmacological dissections of the retinoic acid signaling pathway during mouse embryogenesis. Annu. Rev. Pharmacol. Toxicol. 46, 451-80.
    Nagpal, S. et. al. (1992) RAR-ß4, a retinoic acid receptor isoform is generated from RAR-ß2 by alternative splicing and usage of a CUG initiator codon. Proc. Natl. Acad. Sci. USA. 89, 2718-22.
    Sommer, K.M. et. al. (1999) Elevated retinoic acid receptor ß4 protein in human breast tumor cells with nuclear and cytoplasmic localization. Proc. Natl. Acad. Sci. USA. 96, 8651-6.
Back to top
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net