to see a detailed,interactive version of this map
|
NOTE: PathFinder requires the Flash plug-in for proper viewing. It will launch in a separate pop-up window, please turn off any pop-up blockers for this web site. |
Copyright ©2006 ProteinLounge.com
| |
P53 Signaling
The phosphoprotein p53 is a cell stress sensor molecule. P53 responds to stress factors that damage DNA, such as ionizing radiation, genotoxic drugs and free radicals. In normal cells, p53 functions as a key coordinator of DNA-damage cell-cycle check point activity. It stops DNA replication and facilitates DNA repair. When P53 is activated it drives the expression of proteins involved in DNA repair, cell-cycle arrest, redox regulation, protein degradation and apoptosis. Depending upon the cell type and context, p53 is an effective promoter of apoptosis, primarily, but not exclusively, via activation of mitochondrial apoptotic machinery (Noxa, PUMA, Bax, p53AIP1). The effects of p53 are limited primarily by mouse double minute 2 (mdm2) protein. P53 is activated by various kinases such as casein kinase 2 (CK2/CSNK), DNA-PK or ataxia-telangiecstasia mutated (ATM)-associated protein kinase (Box 1 motif). At least seven negative and three positive feedback loops affecting p53 activity are recognized. Six act through the MDM-2 protein to regulate p53 activity. The p53 circuit communicates with the Wnt-beta-catenin, IGF-1-AKT, Rb-E2F, p38 MAP kinase, cyclin-cdk, p14/19 ARF pathways and the cyclin G-PP2A, and p73 gene products. In addition to MDM-2, there are two other ubiquitin ligases that can regulate p53 in an autoregulatory manner: Cop-1 and Pirh-2.
P53 is recognized as tumor suppressor protein p53 because it plays a critical role in the suppression of tumors exposed to irradiation and genotoxic chemicals. The p53 gene is mutated in over 50% of all human cancers. Most of the mutations in p53 associated with cancers map to the central core sequence specific DNA binding domain or the C-terminal tetramerization domain. Mutations in these domains inhibit the ability of p53 to bind DNA and activate specific genes or to assume a fully competent tetrameric transcription factor structure, respectively. Mutations in P53 can accumulate in cells under oncogenic pressure.
|
References:
- Erster, S. et. al. (2004) In vivo mitochondrial p53 translocation triggers a rapid first wave of cell death in response to DNA damage that can precede p53 target gene activation. Mol. Cell Biol. 24, 6728-6741.
- Kemp, C.J. et. al. (2001) p53 induction and apoptosis in response to radio- and chemotherapy in vivo is tumor-type-dependent. Cancer Res. 61, 327-332.
- Harris, S.L. et. al. (2005) The p53 pathway: positive and negative feedback loops. Oncogene. 24, 2899-2908.
- Prives, C. and Hall, P.A. (1999) The p53 pathway. J. Pathol. 187, 112-126.
- Sionov, R.V. and Haupt, Y. (1999) The cellular response to p53: the decision between life and death. Oncogene. 18, 6145-6157.
|
Back to top |
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net |