to see a detailed, interactive version of this map
|
NOTE: PathFinder requires the Flash plug-in for proper viewing. It will launch in a separate pop-up window, please turn off any pop-up blockers for this web site.
|
Copyright ©2006 ProteinLounge.com
| |
PTEN Pathway
PTEN, phosphatase and tensin homolog (TEP1), is a dual specificity protein/3-lipid phosphatase that functions as a tumor suppressor and negative growth regulator. Inactivation of PTEN is associated with a variety cancers including glioblastoma, melanoma, and prostate, breast, and endometrial cancers.
Under normal conditions, PTEN down-regulates the effects of PI3-kinase activity mediated through the formation of PtdIns(3,4,5) trisphosphate. PTEN (TEP1), as a lipid phosphatase, dephosphorylates phosphatidylinositol 3,4,5-trisphosphate (PtdIns(3,4,5)P3) to phosphatidylinositol 4,5-bisphosphate (PtdIns(4,5)P2) (PIP2), the substrate for PI-phospholipase C. PtdIns(3,4,5)P3 and it’s dephosphorylated product PtdIns(3,4) bisphosphate, formed by 5’-lipid phosphatases, such as Src homology 2 domain containing inositol polyphosphate phosphatase (SHIP), are membrane associated lipids that regulate cell processes that promote survival, growth, mitosis, protein synthesis, and motility. These lipids promote cell membrane association and activation of a variety of kinase and lipases that contain peckstrin homology (PH) domains. A large number of signaling molecules are regulated through PH domains and PtdIns(3,4,5) trisphosphate and PtdIns(3,4) bisphosphate. The specific signals depend upon the cell type and context but generally include members of the following families: phosphoinositide-dependent kinases (PDK1), Akt/PKB, p70S6K, various GEFs, Bruton’s protein kinase (Btk), integrin-linked kinase (ILK), atypical protein kinases C (PKC), centaurin-1, Gab-1, DOS and phospholipase Cgamma.
PTEN, as a protein phosphatase, can dephosphorylate focal adhesion kinase-1 (FAK) and interrupt focal adhesion complex (FAC)-associated growth factor and integrin signaling leading to growth and survival. PTEN dephosphorylation of FAK promotes apoptosis in response to cell detachment from the extracellular matrix (ECM). Consequently, defects in PTEN activity may contribute increased tumor invasiveness.
|