to see a detailed, interactive version of this map
|
NOTE: PathFinder requires the Flash plug-in for proper viewing. It will launch in a separate pop-up window, please turn off any pop-up blockers for this web site.
|
Copyright ©2006 ProteinLounge.com
| |
Rac1 Pathway
Rac1 is a member of a RhoGTPase subfamily (Rac1-Rac4) that transduces extracellular signals from G-coupled protein receptors (GPCR), integrins and growth factor receptors to effector molecules that modulate multiple signaling pathways. While Rac1 supports the activation of MAPkinase pathways and gene activation, it’s most important functions include the regulation of cytoskeletal structures. Together with Rho (regulator of stress fibers) and Cdc42 (regulator of filopodia), Rac modulates the formation of focal adhesion (FA) complexes; membrane ruffles and lamellipodia that contribute to important cell functions related to cell attachment and movement.
Cell movement is a fundamental element of many biological and pathological processes including phagocytosis, wound repair, embryogenesis, inflammatory and immunological responses and tumor metastasis. A critical step in all cell movement is the extension of protrusions in the direction of the desired move. Cells accomplish this by membrane ruffling and the formation of lamellipodia. Rac1 is the primary Rho-GTPase responsible for regulating the cytoskeletal remodeling involved with lamellipodia dynamics.
Activation of these pathways requires the release of Rac1 from Rac1:RhoGDIs complexes in the cytoplasm by Rac1-GDI kinases and targeting to cell membranes by isoprenyl moieties. Membrane associated Rac1 is activated by an extensive array of tissue and location specific guanine nucleotide exchange factors (GEF) such as: Vav-GEFs (Vav1, Vav2 and p95Vav); Swap-70 (and homologue IBP); Kalirin-7 and 8; P-Rex2 and P-Rex2b; betaPIX; Duo, Trio; Fir; FERM; Ras-GRF1; GEFT and Tiam1 which exchanges GTP for Rac1 bound GDP. Some of these GEF are not direct Rac1-GTP exchangers but exchange GTP with other GTPases upstream of Rac1-GTPase activation. Activated Rac1 activates the effectors: PAK-1, -2, -3, -4, MLK3, MRCK, IQGAP, WAVE/IRSp53, FHOS (formin/diaphanous-related protein), PI 5-kinase, BLNK (B cell LiNKer protein) depending upon cell phenotype and context. GEFs provide linkage to specific receptors and downstream effectors of Rac1 signaling pathways. The activity of Rac1 is down-regulated by Rac1-GAPs (GTPase activating proteins).
|
References:
King AJ, Sun H, Diaz B, Barnard D, Miao W, Bagrodia S, Marshall MS: The protein kinase Pak3 positively regulates Raf-1 activity through phosphorylation of serine 338. Nature 1998, 396:180-183.
Frost JA, Steen H, Shapiro P, Lewis T, Ahn N, Shaw PE, Cobb MH: Cross-cascade activation of ERKs and ternary complex factors by Rho family proteins.
EMBO J 1997, 16:6426-6438.
Begum, R. et. al. (2004) The role of Rho GTPases in the regulation of the rearrangement of actin cytoskeleton and cell movement. Exp. Mol. Med. 36, 358-366.
Chan, A. Y. et. al. (2005) Roles of the Rac1 and Rac3 GTPases in human tumor cell invasion. Oncogene. 24, 7821-7829.
Disanza, A. et. al. (2005) Actin polymerization machinery: the finish line of signaling networks, the starting point of cellular movement. Cell Mol. Life Sci. 62, 955-970
Fujiwara, H. et. al. (2004) Rac regulates integrin-mediated endothelial cell adhesion and migration on laminin-8. Exp. Cell Res. 292, 67-77.
Grimsley, C. M. et. al. (2004) Dock180 and ELMO1 proteins cooperate to promote evolutionarily conserved Rac-dependent cell migration. J. Biol. Chem. 279, 6087-6097.
Katoh, H. et. al. (2006) Activation of Rac1 by RhoG regulates cell migration. J. Cell Sci. 119, 56-65.
Kurokawa, K. et. al. (2004) Coactivation of Rac1 and Cdc42 at lamellipodia and membrane ruffles induced by epidermal growth factor. Mol. Biol. Cell. 15, 1003-1010.
Lu, M. and Ravichandran, K. S. (2006) Dock180-ELMO cooperation in Rac activation. Methods Enzymol. 406:388-402.
Steffen, A. et. al. (2004) Sra-1 and Nap1 link Rac to actin assembly driving lamellipodia formation. EMBO J. 23, 749-759.
|
Back to top |
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net
|