Plant Biotechnology
Plant Molecular Biology
Product Lines
Product Highlights
Key Resources
Plant Proteomics
Plant Tissue Culture
Plant Immunochemistry
Lab Supplies & Books
Tissue Culture Protocols
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 SpyLine™ Plant mRNA Capture Kits
Plant Molecular Biology
The streamlined system for measuring plant gene expression
Ordering Information
The SpyLine Plant mRNA Capture Kits are configured three different ways: for mRNA capture with no RT-PCR enzymes provided, for mRNA capture with RT-PCR enzymes provided, and for mRNA capture with SYBR® Green RT-PCR enzymes provided.
Product Number Kit Name
SL0010-1KT SpyLine™ Plant mRNA Capture Kit
sufficient for: 96 mRNA isolations from tissue samples ≤70 mg each
Product Number Kit Name
SL0020-1KT SpyLine™ Plant mRNA Capture Real-Time PCR Kit
sufficient for: 96 mRNA isolations and 96 SYBR Green qRT-PCR reactions
Product Number Kit Name
SL0030-1KT SpyLine™ Plant mRNA RT-PCR Kit
sufficient for: 96 mRNA isolations and 96 RT-PCR reactions
The RT-PCR enzyme mixes that are provided in Product SL0020 and SL0030 have been optimized for use with the SpyLine protocol. However, it is likely that enzyme mixes from quality manufacturers other than Sigma will perform similarly.
Assays that use TaqMan® primers and probes should be performed using SL0010 and Sigma's Quantitative RT-PCR ReadyMix™ (Prod. # D7440).
SpyLine Plant mRNA Capture Kits tell a friend button
  SpyLine Plant mRNA Capture Kits Additional Header
Product Summary
Detailed Product Description
Instruction Manual (61 Kb PDF)