Plant Biotechnology
Plant Molecular Biology
Product Lines
Product Highlights
Key Resources
Plant Proteomics
Plant Tissue Culture
Plant Immunochemistry
Lab Supplies & Books
Tissue Culture Protocols
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Maize SSR Primer Set
Plant Molecular Biology
Using the Maize SSR Primer Set to screen SSR markers for mapping projects:
  •   Reduces the upfront costs of custom oligos synthesis.
  •   Reduces the time and labor involved in compiling primer sequences from SSR databases and
       assembling custom oligo orders
  •   Virtually eliminates human error inherent in diluting and aliquoting large sets of primers.
Product Format
The Maize SSR Primer Set has 384 unique primer pairs contained in four 96-well plates. The primers are shipped lyophilized at 50µM/200µl and are easily reconstituted with TE buffer. Working stocks of 2.5µM are created with a single dilution an additional 96-well PCR plates are included with each set.
Click on plates for primer and sequence information
plate1 plate2 plate3 plate4
View the entire set (68 Kb PDF)
Application Poster (1.5Mb PDF)
Don't need 384 primers? Even if you used only half of the Set, you could still save time and money.
Cost Comparison
Custom oligo pricing fluctuates constantly. However, based on competitive pricing of $0.14 US per base, the Maize SSR Primer Set offers a significant reduction in primer cost.
Order Information
Product Code Product Name Package Size
M4193 Maize SSR Primer Set 1 Set
Also available
Product Code Product Name Package Size
M8818 Maize Polymorphic SSR Primer Set 1 Set
Have any questions about the information on this webpage? Ask Us!
Send this page to a colleague.
Sample Data
Screening for Polymorphic SSRs
Figure 1. Eight primer pairs from the Set were used to screen SSR markers in two parental lines (P1 and P2) and the hybrid (F1).