Plant Biotechnology
Plant Molecular Biology
Product Lines
Product Highlights
Key Resources
Plant Proteomics
Plant Tissue Culture
Plant Immunochemistry
Lab Supplies & Books
Tissue Culture Protocols
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Maize SSR Primer Sequence
Plant Molecular Biology
Maize SSR Home
Plate 1
Plate 2 (Searchable Format - 29 Kb PDF)
   Row A   Row B   Row C   Row D   Row E   Row F   Row G   Row H
Plate 3
Plate 4
Plate 2 — Click on Gene Names to view maizegdb.org
Row A     back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc1736 2/A1 2 627.98
p-bnlg469 2/A2 2 650.1
p-umc2184 2/A3 2 692.4
p-phi101049 2/A4 2 712.1
p-umc2214 2/A5 2 728.36
p-umc2118 2/A6 3 0
p-umc1780 2/A7 3 11.2
p-umc2256 2/A8 3 23.4
p-umc2049 2/A9 3 38.7
p-umc2377 2/A10 3 47.91
p-umc1458 2/A11 3 67.2
p-umc1886 2/A12 3 78.5
Row B back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc1814 2/B1 3 86.84
p-bnlg1647 2/B2 3 103.3
p-umc2258 2/B3 3 127.8
p-umc2259 2/B4 3 131.7
p-phi036 2/B5 3 159
p-umc2158 2/B6 3 176.6
p-umc1655 2/B7 3 191.1
p-mmc0132 2/B8 3 208.6
p-umc2263 2/B9 3 227.8
p-umc1223 2/B10 3 234.4
p-umc1968 2/B11 3 250.4
p-umc1683 2/B12 3 266
Row C back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc1773 2/C1 3 280.4
p-phi053 2/C2 3 296.1
p-bnlg1601 2/C3 3 306.1
p-umc1501 2/C4 3 325.4
p-umc1307 2/C5 3 339.61
p-umc2265 2/C6 3 354
p-umc1973 2/C7 3 371.4
p-umc1539 2/C8 3 390.3
p-umc1876 2/C9 3 414.69
p-umc2266 2/C10 3 434.3
p-bnlg2241 2/C11 3 454.31
p-umc1644 2/C12 3 473.1
Row D back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-bnlg1160 2/D1 3 491.4
p-bnlg197 2/D2 3 511.5
p-umc2050 2/D3 3 538.2
p-umc1148 2/D4 3 552.14
p-umc1404 2/D5 3 568.3
p-umc1825 2/D6 3 579.5
p-umc2274 2/D7 3 608.6
p-umc2081 2/D8 3 627.1
p-umc2276 2/D9 3 652.4
p-umc2174 2/D10 3 683.6
p-umc2152 2/D11 3 738.7
p-umc2277 2/D12 3 747.5
Row E back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-bnlg1496 2/E1 3 760.9
p-umc1361 2/E2 3 786.39
p-umc1641 2/E3 3 806.9
p-umc1010 2/E4 3 827.4
p-umc2048 2/E5 3 858.62
p-umc1008 2/E6 4 2.9
p-umc2279 2/E7 4 22.9
p-umc1682 2/E8 4 47.6
p-umc2409 2/E9 4 70.05
p-umc2410 2/E10 4 91.57
p-umc1294 2/E11 4 108.43
p-bnlg1126 2/E12 4 135.3
Row F back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc2281 2/F1 4 158.6
p-umc1902 2/F2 4 181.4
p-umc2039 2/F3 4 196.4
p-umc1117 2/F4 4 218.5
p-bnlg490 2/F5 4 237.8
p-umc2282 2/F6 4 254.9
p-umc2282 2/F7 4 271.4
p-umc1175 2/F8 4 287.3
p-umc1702 2/F9 4 305.2
p-mmc0371 2/F10 4 331.3
p-mmc0371 2/F11 4 349.8
p-dupssr16 2/F12 4 371.71
Row G back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-bnlg2291 2/G1 4 392.2
p-umc2038 2/G2 4 411.3
p-umc2038 2/G3 4 428
p-umc1775 2/G4 4 443.2
p-bnlg1444 2/G5 4 462.5
p-bnlg2162 2/G6 4 475.7
p-umc2041 2/G7 4 483.93
p-umc1086 2/G8 4 500.59
p-umc2285 2/G9 4 514.9
p-umc2187 2/G10 4 531.7
p-umc2286 2/G11 4 553.7
p-umc2139 2/G12 4 574.8
Row H back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc1854 2/H1 4 595.7
p-umc1328 2/H2 4 618.1
p-umc1573 2/H3 4 644.3
p-bnlg589 2/H4 4 670.2
p-umc1109 2/H5 4 687.8
p-umc2289 2/H6 4 707.8
p-umc1738 2/H7 4 720.48
p-umc2290 2/H8 4 736.7
p-bnlg1890 2/H9 4 750.2
p-umc1308 2/H10 5 6.3
p-umc2291 2/H11 5 23.3
p-umc1097 2/H12 5 40.8