Plant Biotechnology
Plant Molecular Biology
Product Lines
Product Highlights
Key Resources
Plant Proteomics
Plant Tissue Culture
Plant Immunochemistry
Lab Supplies & Books
Tissue Culture Protocols
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Maize SSR Primer Sequence
Plant Molecular Biology
Maize SSR Home
Plate 1
Plate 2
Plate 3 (Searchable Format - 29 Kb PDF)
   Row A   Row B   Row C   Row D   Row E   Row F   Row G   Row H
Plate 4
Plate 3 — Click on Gene Names to view maizegdb.org
Row A back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc1325 3/A1 5 54.6
p-umc1679 3/A2 5 71.5
p-umc1365 3/A3 5 97.95
p-umc2036 3/A4 5 124.7
p-bnlg565 3/A5 5 150.9
p-bnlg1879 3/A6 5 189.8
p-umc1686 3/A7 5 210.3
p-mmc0351 3/A8 5 230.4
p-umc2294 3/A9 5 245.5
p-umc1830 3/A10 5 266.3
p-umc1429 3/A11 5 286.6
p-umc2298 3/A12 5 307
Row B back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-bnlg2323 3/B1 5 328.5
p-umc1349 3/B2 5 346.5
p-umc1221 3/B3 5 368.4
p-mmc0081 3/B4 5 389.9
p-umc1155 3/B5 5 410.8
p-umc2304 3/B6 5 428.3
p-bnlg1237 3/B7 5 454.01
p-umc1019 3/B8 5 469.6
p-umc1941 3/B9 5 492.6
p-umc2216 3/B10 5 518.37
p-phi085 3/B11 5 528.7
p-umc1537 3/B12 5 553.61
Row C back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc1375 3/C1 5 571.66
p-bnlg1118 3/C2 5 590.4
p-bnlg118 3/C3 5 609.4
p-umc1792 3/C4 5 625.8
p-umc1225 3/C5 5 641.4
p-bnlg389 3/C6 5 661.56
p-umc1153 3/C7 5 676.7
p-umc1002 3/C8 6 9.1
p-umc2309 3/C9 6 27.8
p-umc2068 3/C10 6 47.8
p-umc1753 3/C11 6 70.37
p-umc2313 3/C12 6 91.9
Row D back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc1186 3/D1 6 114.25
p-y1SSR 3/D2 6 120.5
p-umc1257 3/D3 6 145.7
p-umc1887 3/D4 6 166.8
p-umc1796 3/D5 6 189.5
p-nc010 3/D6 6 211.5
p-umc2006 3/D7 6 228.9
p-umc2318 3/D8 6 244.7
p-umc1250 3/D9 6 254.5
p-umc1826 3/D10 6 267.7
p-umc1314 3/D11 6 284.3
p-umc1379 3/D12 6 297.1
Row E back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-bnlg1174 3/E1 6 315.4
p-umc1020 3/E2 6 325.9
p-umc2065 3/E3 6 335.71
p-umc1805 3/E4 6 346.22
p-bnlg1732 3/E5 6 373.8
p-umc1912 3/E6 6 388.7
p-umc2322 3/E7 6 404.4
p-umc1520 3/E8 6 414.48
p-umc2170 3/E9 6 435.1
p-umc2170 3/E10 6 452.7
p-umc1490 3/E11 6 466.5
p-umc2323 3/E12 6 483.5
Row F back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc2165 3/F1 6 502.9
p-bnlg1740 3/F2 6 510.6
p-bnlg1740 3/F3 6 531.8
p-umc2324 3/F4 6 544.5
p-umc2177 3/F5 7 0
p-umc1241 3/F6 7 13.8
p-umc1642 3/F7 7 27.2
p-umc1672 3/F8 7 43.8
p-bnlg2132 3/F9 7 53.3
p-umc1840 3/F10 7 70.67
p-umc1159 3/F11 7 92
p-umc1159 3/F12 7 110.59
Row G back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc2325 3/G1 7 127.6
p-umc1401 3/G2 7 153
p-umc1927 3/G3 7 168.5
p-umc1879 3/G4 7 183.4
p-umc1932 3/G5 7 204.8
p-umc1339 3/G6 7 214.12
p-umc1983 3/G7 7 244.3
p-umc1393 3/G8 7 258.4
p-bnlg657 3/G9 7 276.79
p-mmc0411 3/G10 7 292.7
p-umc1450 3/G11 7 315.9
p-phi091 3/G12 7 330.6
Row H back to top
Gene Name Plate location Linkage group (chrom.) Map location (IBMn)
p-umc1660 3/H1 7 351.4
p-umc1837 3/H2 7 368.9
p-bnlg2271 3/H3 7 383.8
p-umc1936 3/H4 7 405.5
p-umc1301 3/H5 7 405.5
p-bnlg1666 3/H6 7 430.5
p-umc1029 3/H7 7 444.7
p-umc1543 3/H8 7 457.44
p-umc1543 3/H9 7 472.6
p-bnlg2259 3/H10 7 489.2
p-umc1412 3/H11 7 518.9
p-phi069 3/H12 7 545.2