Plant Biotechnology
Plant Molecular Biology
Plant Proteomics
Plant Tissue Culture
Plant Immunochemistry
Lab Supplies & Books
Tissue Culture Protocols
Media Preparation
Vitamin Mixtures
Growth Regulators
Iron Chelate Solution
Gelling Agents
Media Sterilization
Antibiotics
Classic Plant Media
Murashige & Skoog
Orchid Culture Media
Pathology Media
Phycology Media
Explant Sterilization
Silver Thiosulfate
Sunbag Vessels
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
Classic Plant Media
Composition
Plant Biotech letterhead
  C 1416
D 6162
G 5768
G 5893
H 2395
M 6774
M 5524
Q 3376
S 6765
W 0876
Chu (N6) basal salt mixture
DKW/Juglans basal salt mixture
Gamborg's B-5 basal salt mixture
Gamborg's B-5 basal salt mixture with minimal organics
Hoagland's No. 2 basal salt mixture
McCown's woody plant basal salt mixture
Murashige and Skoog basal salt mixture (MS)
Quoirin and Lepoivre basal salt mixture
Schenk and Hildebrandt basal salt mixture
White's basal salt mixture
Component (mg/L) C 1416 D 6162 G 5768 G 5893 H 2395 M 6774 M 5524 Q 3376 S 6765 W 0876
Aluminum chloride • 6H2O                    
Ammonium nitrate   1416.0       400.0 1650.0 400.0    
Ammonium phosphate monobasic         115.03       300.0  
Ammonium sulfate 463.0   134.0 134.0            
Boric acid 1.6 4.8 3.0 3.0 2.86 6.2 6.2 6.2 5.0 1.5
Calcium chloride anhydrous 125.33 112.5 113.24 113.24   72.5 332.2   151.0  
Calcium nitrate   1367.0     656.4 386.0   833.77   200.0
Calcium phosphate tribasic                    
Cobalt chloride • 6H2O     0.025 0.025     0.025 0.025 0.1  
Cupric sulfate • 5H2O   0.25 0.025 0.025 0.08 0.25 0.025 0.025 0.2  
Na2-EDTA 37.25 45.4 37.3 37.25   37.3 37.26 37.3 20.0  
ZnNa2-EDTA                    
Ferric chloride • 6H2O                    
Ferric citrate                    
Ferric sulfate                   2.5
Ferric tartrate • 2H2O         5.32          
Ferrous sulfate • 7H2O 27.85 33.8 27.8 27.85     27.8 27.8 15.0  
Magnesium sulfate 90.37 361.49 122.09 122.09 240.76 180.7 180.7 175.79 195.4 360.0
Manganese chloride • 4H2O         1.81          
Manganese sulfate • H2O 3.33 33.5 10.0 10.0   22.3 16.9 0.76 10.0 5.04
Molybdenum trioxide         0.016          
Molybdic acid (sodium salt) • 2H2O   0.39 0.25 0.25   0.25 0.25 0.25 0.1  
Nickel chloride • 6H2O                    
Nickel sulfate • 6H2O   0.005                
Potassium chloride                   65.0
Potassium iodide 0.8   0.75 0.75     0.83 0.08 1.0 0.75
Potassium nitrate 2830.0   2500.0 2500.0 606.6   1900.0 1800.0 2500.0 80.0
Potassium phosphate monobasic 400.0 265.0       170.0 170.0 270.0    
Potassium sulfate   1559.0       990.0        
Sodium nitrate                    
Sodium phosphate monobasic     130.5 130.5           16.5
Sodium sulfate                   200.0
Zinc nitrate • 6H2O   17.0                
Zinc sulfate • 7H2O 1.5   2.0 2.0 0.22 8.6 8.6 8.6 1.0 2.67
Organics (mg/L) C 1416 D 6162 G 5768 G 5893 H 2395 M 6774 M 5524 Q 3376 S 6765 W 0876
Activated charcoal                    
Adenine hemisulfate                    
Agar                    
6-Benzylaminopurine (BA)                    
D-Biotin                    
6gamma-gamma-
Dimethylallylaminopurine
Glycine (free base)                    
Indole-3-acetic acid                    
Indole-3-butyric acid                    
Kinetin                    
MES (free acid)                    
myo-Inositol       100.0            
alpha-Naphthaleneacetic acid                    
Nictotinic acid (free acid)       1.0            
Peptone                    
Pyridoxine • HCl       1.0            
Sucrose                    
Thiamine • HCl       10.0            
  C 1416 D 6162 G 5768 G 5893 H 2395 M 6774 M 5524 Q 3376 S 6765 W 0876
Grams of powder to prepare 1 L 4.0 5.2 3.1 3.2 1.6 2.3 4.3 3.6 3.2 0.934
pH 1 0.5 at room temperature 4.1 4.1 4.0 4.0 4.7 3.9 3.9 3.9 4.2 4.7
Back to Top