Plant Biotechnology
Plant Molecular Biology
Plant Proteomics
Plant Tissue Culture
Plant Immunochemistry
Lab Supplies & Books
Tissue Culture Protocols
Media Preparation
Vitamin Mixtures
Growth Regulators
Iron Chelate Solution
Gelling Agents
Media Sterilization
Antibiotics
Classic Plant Media
Murashige & Skoog
Orchid Culture Media
Pathology Media
Phycology Media
Explant Sterilization
Silver Thiosulfate
Sunbag Vessels
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
Iron Chelate Solution
Preparation and Use
Plant Biotech letterhead
To prepare 1L of a 10 X iron chelate solution:
  1. Dissolve 0.278 g of ferrous sulfate heptahydrate (Product No. F 8263) in 350 ml of tissue culture grade water (Product No. W 3500). Heating may be required.
  2. Dissolve 0.337 g of disodium EDTA (Product No. E 6635) in 350 ml using tissue culture grade water. Heating may be required.
  3. When both components are dissolved, combine the solutions and bring up to the final volume of 1 L using tissue culture grade water. This will yield a clear yellow solution. (Concentrations of components are based upon those present in MS basal salt formulations and utilized in McCown's Woody Plant Medium).
ALTERNATE METHOD
To prepare 1L of a 200 X iron chelate stock solution:
  1. Dissolve 7.45 g of disodium EDTA (Product No. E 6635) in 900 ml of tissue culture grade water. A clear solution is obtained in 20 minutes at room temperature (25-27 °C).
  2. Gradually add 5.52 g of ferrous sulfate heptahydrate (Product No. F 8263). A clear yellow solution is obtained immediately.
  3. Store the iron chelate stock solution in an amber bottle to protect it from exposure to light.
  4. Use 5 ml of the 200 X iron chelate stock solution to prepare 1 L of culture medium requiring 27.8 mg/L of ferrous sulfate and 37.25 mg/L of disodium salt of EDTA (e.g., Murashige and Skoog, 1962; Gamborg, et al., 1968; Lloyd and McCown, 1981).