Plant Biotechnology
Plant Molecular Biology
Plant Proteomics
Plant Tissue Culture
Plant Immunochemistry
Lab Supplies & Books
Tissue Culture Protocols
Media Preparation
Vitamin Mixtures
Growth Regulators
Iron Chelate Solution
Gelling Agents
Media Sterilization
Antibiotics
Classic Plant Media
Murashige & Skoog
Orchid Culture Media
Pathology Media
Phycology Media
Explant Sterilization
Silver Thiosulfate
Sunbag Vessels
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
Sunbag Vessels
Description and Use
Plant Biotech letterhead
Sunbag (Product No. B 7026)
    The Sunbag offered by Sigma was originally developed for the culture of mushrooms, but may be adapted for the culture of microshoots. The bag has a thickness of 55 5m and measures 320 x 450 mm. This is a gusset bag with a square bottom, so it stands upright. The bag has a 0.3 5m membrane measuring 36 x 36 mm, which allows for gas exchange.
    To use the bag for plant cell culture, fill the bottom of the bag with an appropriate amount of medium (e.g. 500-1000 ml). Fold over the top of the bag and hold in place with either a paper clip or clothespin. Autoclave the media in the bag and then allow to cool on a flat surface. DO NOT AUTOCLAVE BAGS WITHOUT MEDIA AS THEY WILL HARDEN AND BE UNUSABLE. Once the media has solidified, the bag can be opened under sterile conditions and the explants placed on the culture media. The bag is then resealed using clothespins or paper clips or sealed with a plastic heat sealer.