PathFinder
Origami contest Antibodies Extract-N-Amp (PCR) Prestige Antibodies
(powered by Atlas)
Universal Proteomics Standard Look Closer at the Shape of Life Science.
Origami Goose
ORIGAMI CONTEST
Our Challenge: Master the ancient art of Origami, and out-fold your fellow scientists!
Enter to Win!
Picture Frame
IT'S AN ORIGINAL
To receive your FREE copy of the 2007-2008 Catalog,
click here.
See Your Proteomic Discovey Take Flight!
Align the Course of Your MS Strategy
Sigma’s complex Proteomics Standards are the leading-edge tools for aligning proteomic strategies. Developed in collaboration with the Association of Biomolecular Resource Facilities (ABRF), Proteomics Standards Research Group (sPRG), the original Universal Proteomics Standard and the enhanced Proteomics Dynamic Range Standard allow you to:
    Optimize your discovery protocol
    Confirm system suitability
    Compare analytical results day-to-day or lab-to-lab
Image of Red Cube Universal Proteomics Standard, UPS1
Developed in collaboration with the Association of Biomolecular Resource Facilities Proteomics Standards Research Group (sPRG), the Universal Proteomics Standard (UPS1) contains 48 human proteins (5 pmols of each) ranging in molecular weight from 6,000 to 83,000 daltons.
Go to Universal Proteomics Standard, UPS1
Image of Red Cube Proteomics Dynamic Range Standard, UPS2
This new standard is an enhancement of Sigma's original Universal Proteomics Standard (UPS1). The same complex mixture of 48 human proteins has been formulated into a dynamic range of concentrations, ranging from 500 amoles to 50 pmoles.
Go to Proteomics Dynamic Range Standard, UPS2
Back to Top