|
NEW Proteomics Dynamic Range Standard, UPS2
- Troubleshoot and optimize your analytical protocol
- Confirm system suitability before analyzing critical samples
- Normalize analytical results day-to-day or lab-to-lab
- Determine your limit of detection
|
|
This new standard is an enhancement of Sigma's original Universal Proteomics Standard (UPS1). The same complex mixture of 48 human proteins has been formulated into a dynamic range of concentrations, ranging from 0.5 fmoles to 50 pmoles.
UPS1 | UPS2 | List of UPS Proteins | References | 2006 ABRF Study | UPS1 & UPS2 Home |
| Ordering Information |
| Product # |
Product Name |
Pkg Size |
Add to Cart |
| UPS2 |
Proteomics Dynamic Range Standard |
1 set |
|
Each set contains one vial of Universal Proteomics Standard and one vial (20 µg) of Proteomics Grade Trypsin (T6567).
The Proteomics Dynamic Range Standard contains the following proteins:
| Protein Name [Synonym] |
MW (Da) (calculated) |
Amount (fmoles) |
Protein Name [Synonym] |
MW (Da) (calculated) |
Amount (fmoles) |
| Carbonic anhydrase 1 |
28,738 |
50000 |
Antithrombin-III |
49,033 |
50 |
| Carbonic anhydrase 2 |
29,095 |
50000 |
Beta-2-microglobulin |
11,729 |
50 |
Complement C5 [Complement C5a] |
8,266 |
50000 |
BH3 interacting dom1ain death agonist [BID] |
21,978 |
50 |
| Hemoglobin alpha chain |
15,127 |
50000 |
Cathepsin D |
26,624 |
50 |
| Hemoglobin beta chain |
15,867 |
50000 |
Glutathione S-transferase A1 [GST A1-1] |
25,482 |
50 |
| Leptin |
16,024 |
50000 |
Insulin-like growth factor II |
7,464 |
50 |
| Serum albumin |
66,393 |
50000 |
Platelet-derived growth factor B chain |
12,286 |
50 |
| Ubiquitin |
9,387 |
50000 |
Thioredoxin |
12,424 |
50 |
| Catalase |
59,583 |
5000 |
Cathepsin G |
26,751 |
5 |
| Cytochrome b5 |
16,021 |
5000 |
Cytochrome c [Apocytochrome c] |
11,608 |
5 |
| Epidermal growth factor |
6,211 |
5000 |
Gelsolin |
82,954 |
5 |
| Myoglobin |
17,051 |
5000 |
Glutathione S-transferase P [GST] |
23,220 |
5 |
| NAD(P)H dehydrogenase [quinone] 1 [DT Diaphorase] |
30,984 |
5000 |
GTPase HRas [Ras protein] |
21,292 |
5 |
| Peptidyl-prolyl cis-trans isomerase A [Cyclophilin A] |
17,947 |
5000 |
Serotransferrin [Apotransferrin] |
75,143 |
5 |
| Peroxiredoxin 1 |
22,106 |
5000 |
Ubiquitin-conjugating enzyme E2 C [UbcH10] |
20,473 |
5 |
| Small ubiquitin-related modifier 1 [SUMO-1] |
37,420 |
5000 |
Ubiquitin-conjugating enzyme E2 E1 [UbcH6] |
22,222 |
5 |
| Alpha-lactalbumin |
14,070 |
500 |
Annexin A5 |
35,782 |
0.5 |
Creatine kinase M-type [CK-MM] |
43,070 |
500 |
C-reactive protein |
23,030 |
0.5 |
| Histidyl-tRNA synthetase [Jo-1] |
58,223 |
500 |
Fatty acid-binding protein |
14,716 |
0.5 |
| Lysozyme C |
14,692 |
500 |
Interleukin-8 |
8,381 |
0.5 |
| Neddylin [Nedd8] |
9,071 |
500 |
Lactotransferrin |
78,289 |
0.5 |
| Retinol-binding protein |
21,065 |
500 |
Microtubule-associated protein tau [Tau protein] |
46,810 |
0.5 |
| Ribosyldihydronicotinamide dehydrogenase (quinone) [Quinone oxidoreductase 2 or NQO2] |
25,817 |
500 |
Superoxide dismutase [Cu-Zn] |
15,800 |
0.5 |
| Ubiquitin-conjugating enzyme E2 I [UbcH9] |
22,907 |
500 |
Tumor necrosis factor [TNF-alpha] |
17,350 |
0.5 |
back to top
back to Proteomics & Protein Expression homepage
|