|
Sigma now offers both the Universal Proteomics Standard and the Proteomics Dynamic range Standard as complex, well-defined, well characterized reference standards for mass spectrometry. Both standards contain the same 48 human proteins ranging in molecular mass from 6,000 to 83,000 Daltons. Each constituent protein has been HPLC purified and AAA quantitated prior to formulation.
UPS1 | UPS2 | List of UPS Proteins | References | 2006 ABRF Study | UPS1 & UPS2 Home |
 | Troubleshoot and optimize your analytical protocol |
 | Confirm system suitability before analyzing critical samples |
 | Normalize analytical results day-to-day or lab-to-lab |
 | Determine your limit of detection |
Universal Proteomics Standard, UPS1
 |
Developed in collaboration with the Association of Biomolecular Resource
Facilities Proteomics Standards Research Group (sPRG), the Universal Proteomics Standard (UPS1) contains 48 human proteins (5 pmols of each) ranging in molecular weight from 6,000 to 83,000 daltons.
|
NEW Proteomics Dynamic Range Standard, UPS2
 |
This new standard is an enhancement of Sigma's original Universal Proteomics
Standard (UPS1). The same complex mixture of 48 human proteins has been formulated into a dynamic range of concentrations, ranging from 500 amoles to 50 pmoles.
|
| Ordering Information |
| Product # |
Product Name |
Pkg Size |
Add to Cart |
| UPS1 |
Universal Proteomics Standard |
1 set |
|
| UPS2 |
Proteomics Dynamic Range Standard |
1 set |
|
Each set contains one vial of Universal Proteomics Standard and one vial (20 µg) of Proteomics Grade Trypsin (T6567).
References
The ABRF sPRG 2006 Study
In the Fall and Winter of 2005/2006, the ABRF sPRG (Proteomics Standards Research Group) conducted a study to assess the analytical capabilities of proteomics laboratories. Approximately 125 labs from across the world volunteered to participate. Each lab received a complex mixture of 49 unknown proteins and were asked to identify as many of these proteins as possible using their best analytical strategies. The results, presented in February 2006, were quite impressive and in some cases, surprising. Learn more about the sPRG’s 2006 study.
List of UPS Proteins
| UniProt Accession Number |
UPS1 Amount (fmol) |
UPS2 Amount (fmol) |
UniProt Protein Name [Synonym] |
MW (Da) (calculated) |
Source or recombinant |
Host |
Tag |
Potential PTMs |
| P00915 |
5,000 |
50,000 |
Carbonic anhydrase 1 |
28,738 |
Erythrocytes |
|
|
Acetylation |
| P00918 |
5,000 |
50,000 |
Carbonic anhydrase 2 |
29,095 |
Erythrocytes |
|
|
Acetylation |
| P01031 |
5,000 |
50,000 |
Complement C5 [Complement C5a] |
8,266 |
Recombinant |
E. coli |
|
|
| P69905 |
5,000 |
50,000 |
Hemoglobin alpha chain |
15,127 |
Erythrocytes |
|
|
|
| P68871 |
5,000 |
50,000 |
Hemoglobin beta chain |
15,867 |
Erythrocytes |
|
|
Acetylation, nitrosylation, glycosylation |
| P41159 |
5,000 |
50,000 |
Leptin |
16,024 |
Recombinant |
E. coli |
|
|
| P02768 |
5,000 |
50,000 |
Serum Albumin |
66,393 |
Recombinant |
Pichia pastoris |
|
|
| P62988 |
5,000 |
50,000 |
Ubiquitin |
9,387 |
Recombinant |
E. coli |
6-His |
|
| P04040 |
5,000 |
5,000 |
Catalase |
59,583 |
Erythrocytes |
|
|
|
| P00167 |
5,000 |
5,000 |
Cytochrome b5 |
16,021 |
Recombinant |
E. coli |
6-His |
|
| P01133 |
5,000 |
5,000 |
Epidermal Growth Factor |
6,211 |
Recombinant |
E. coli |
|
|
| P02144 |
5,000 |
5,000 |
Myoglobin C |
17,051 |
Heart |
|
|
|
| P15559 |
5,000 |
5,000 |
NAD(P)H dehydrogenase [quinone] 1 [DT Diaphorase] C |
30,984 |
Recombinant |
E. coli |
|
|
| P62937 |
5,000 |
5,000 |
Peptidyl-prolyl cis-trans isomerase A [Cyclophilin A] |
17,947 |
Recombinant |
E. coli |
|
|
| Q06830 |
5,000 |
5,000 |
Peroxiredoxin 1 |
22,106 |
Recombinant |
E. coli |
|
|
| P63165 |
5,000 |
5,000 |
Small ubiquitin-related modifier 1 [SUMO-1] |
37,420 |
Recombinant |
E. coli |
GST |
|
| P00709 |
5,000 |
500 |
Alpha-lactalbumin |
14,070 |
Milk |
|
|
Glycosylation |
| P06732 |
5,000 |
500 |
Creatine kinase M-type [CK-MM] |
43,070 |
Heart |
|
|
|
| P12081 |
5,000 |
500 |
Histidyl-tRNA synthetase [Jo-1] |
58,223 |
Recombinant |
E. coli |
|
|
| P61626 |
5,000 |
500 |
Lysozyme C |
14,692 |
Milk |
|
|
|
| Q15843 |
5,000 |
500 |
Neddylin [Nedd8] |
9,071 |
Recombinant |
E. coli |
|
|
| P02753 |
5,000 |
500 |
Retinol-binding protein |
21,065 |
Urine |
|
|
|
| P16083 |
5,000 |
500 |
Ribosyldihydronicotinamide dehydrogenase [quinone] [Quinone oxidoreductase 2] [NQO2] |
25,817 |
Recombinant |
E. coli |
|
|
| P63279 |
5,000 |
500 |
Ubiquitin-conjugating enzyme E2 I [UbcH9] |
22,907 |
Recombinant |
E. coli |
|
|
| P01008 |
5,000 |
50 |
Antithrombin-III |
49,033 |
Plasma |
|
|
Glycosylation |
| P61769 |
5,000 |
50 |
Beta-2-microglobulin |
11,729 |
Urine |
|
|
|
| P55957 |
5,000 |
50 |
BH3 Interacting domain death agonist [BID] |
21,978 |
Recombinant |
E. coli |
|
|
| P07339 |
5,000 |
50 |
Cathepsin D |
26,624 |
Liver |
|
|
Glycosylation |
| P08263 |
5,000 |
50 |
Glutathione S-transferase A1 [GST A1-1] |
25,482 |
Recombinant |
E. coli |
|
|
| P01344 |
5,000 |
50 |
Insulin-like growth factor II |
7,464 |
Recombinant |
E. coli |
|
|
| P01127 |
5,000 |
50 |
Platelet-derived growth factor B chain |
12,286 |
Recombinant |
E. coli |
|
|
| P10599 |
5,000 |
50 |
Thioredoxin |
12,424 |
Recombinant |
E. coli |
6-His |
|
| P08311 |
5,000 |
5 |
Cathepsin G |
26,751 |
Sputum |
|
|
Glycosylation |
| P99999 |
5,000 |
5 |
Cytochrome c[Apocytochrome c] |
11,608 |
Recombinant |
E. coli |
|
|
| P06396 |
5,000 |
5 |
Gelsolin |
82,954 |
Plasma |
|
|
Phosphorylation |
| P09211 |
5,000 |
5 |
Glutathione S-transferase P [GST] |
23,220 |
Placenta |
|
|
|
| P01112 |
5,000 |
5 |
GTPase HRas [Ras protein] |
21,292 |
Recombinant |
E. coli |
|
|
| P02787 |
5,000 |
5 |
Serotransferrin [Apotransferrin] |
75,143 |
Plasma |
|
|
Glycosylation |
| O00762 |
5,000 |
5 |
Ubiquitin-conjugating enzyme E2 C [UbcH10] |
20,473 |
Recombinant |
E. coli |
6-His |
|
| P51965 |
5,000 |
5 |
Ubiquitin-conjugating enzyme E2 E1 [UbcH6] |
22,222 |
Recombinant |
E. coli |
6-His |
|
| P08758 |
5,000 |
0.5 |
Annexin A 5 |
35,782 |
Placenta |
|
|
Acetylation |
| P02741 |
5,000 |
0.5 |
C-reactive protein |
23,030 |
Plasma |
|
|
|
| P05413 |
5,000 |
0.5 |
Fatty acid-binding protein |
14,716 |
Plasma |
|
|
Acetylation, phosphorylation |
| P10145 |
5,000 |
0.5 |
Interleukin-8 |
8,381 |
Recombinant |
E. coli |
|
|
| P02788 |
5,000 |
0.5 |
Lactotransferrin |
78,289 |
Milk |
|
|
Glycosylation |
| P10636 |
5,000 |
0.5 |
Microtubule-associated protein tau [Tau protein] |
46,810 |
Recombinant |
E. coli |
6-His |
|
| P00441 |
5,000 |
0.5 |
Superoxide dismutase [Cu-Zn] |
15,800 |
Erythrocytes |
|
|
Acetylation |
| P01343 |
5,000 |
0.5 |
Insulin-like growth factor IA |
7,643 |
Recombinant |
E. coli |
|
|
| P01375 |
5,000 |
0.5 |
Tumor necrosis factor [TNF-alpha] |
17,350 |
Recombinant |
E. coli |
|
|
back to top
back to Proteomics & Protein Expression homepage
|