|
Crystallization Kits |
Crystallization Plates |
Membrane Protein Kit |
Optimization Kits |
Optimization Reagents |
Renaturation Kit
Crystallization Plates
Greiner CrystalQuick Sitting Drop Protein Crystallization Plates
Each reservoir well has an elevated platform with 3 small wells for sample solution, to enable experiments with multiple samples for each reservoir. The bottom of each sample well is optically clear for microscopic examination. Polystyrene VIEWseal transparent seal recommended for sealing these plates. These plates may also be used for hanging drop crystallization by addition the CrystalDrop lid below. The CrystalQuick Plate is designed for automated high throughput crystallography and subsequent X-ray diffraction at reduced costs.
| Product Name |
Reservoir wells |
Sample wells |
Pack quantity |
Greiner Mfr. # |
Product # |
| Greiner CrystalQuick 96-well plate for protein crystallization |
96 x 320µl flat-bottom |
288 x 4µl square |
40 |
609101 |
Z617644 |
| Greiner CrystalQuick 96-well plate for protein crystallization |
96 x 320µl flat-bottom |
288 x 2µl round |
40 |
609120 |
Z666130 |
| VIEWseal transparent seal |
|
|
100 |
676070 |
Z617571 |
back to top
Greiner CrystalDrop Lid for Hanging Drop Protein Crystallization
Designed to give 2 hanging drop positions in each of the 96 wells, the CrystalDrop lid in combination with CrystalQuick plates enables up to 3 sitting and 2 hanging drop experiments to be conducted at each reservoir position. Channels around each of the 96 positions for individual sealing of each well with silicone grease polystyrene.
CrystalDrop is well suited for automated liquid delivery, lid handling and bringing up without drop movement and cross-contamination.
| Product Name |
Pack quantity |
Greiner Mfr. # |
Product # |
| CrystalDrop Lid |
200 |
609150 |
Z617652 |
back to top
Greiner 96 well IMP@CT Plate for Microbatch Protein Crystallization
Developed for crystallization under oil, the conical well wall profile assists placement of sample in centre of the well, while 2mm diameter flat bottom enables microscopic examination. The IMP@CT Plate is designed for automated high throughput crystallography and subsequent x-ray diffraction at reduced costs.
| Product Name |
Pack quantity |
Greiner Mfr. # |
Product # |
| 96 x 19µl wells, 5mm diam. top, 2mm diam. flat bottom |
80 |
673101 |
Z617660 |
| Lid for IMP@CT plate |
80 |
656190 |
L0913 |
back to top
Corning CrystalEX Sitting Drop Protein Crystallization Plates
Manufactured from an advanced, optically clear copolymer that has better chemical resistance, lower water absorption and lower water permeability than Polystyrene, Corning plates reduce protein or reagent losses that can significantly change concentration of protein solutions, particularly important for 384 (192 sample + 192 reservoir wells) format plate. This advanced polymer also provides high optical clarity with minimum background interference when viewing crystals under polarized light.
| Product Name |
Reservoir wells |
Sample wells |
Pack quantity |
Greiner Mfr. # |
Product # |
| Corning Protein Crystallization Plate |
96 x 210µl |
96 x 10µl round 3mm diam., conical bottom |
50 |
3773 |
Z709409 |
| Corning Protein Crystallization Plate |
192 x 100µl |
192 x 3µl round, 2.3mm diam., flat bottom |
50 |
3775 |
Z709417 |
| Universal optical sealing tape |
|
|
100 |
6575 |
Z709425 |