Proteomics and Protein Expr.
Protein Expression
Protein Analysis
Protein Quantitation
Protein Staining/Detection
Protein Electrophoresis
Western Blotting
Mass Spectrometry
X-Ray Crystallography
Chromatography
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Product Lines
X-Ray Crystallography
Crystallization Kits | Crystallization Plates | Membrane Protein Kit | Optimization Kits | Optimization Reagents | Renaturation Kit
Optimization Kits
Additive Kit
Detergent Kit
Additive Kit
Further optimization of preliminary crystallization conditions is often achieved by addition of traces of small molecules. Although the mechanisms are often not investigated or understood, additives seem to manipulate sample-solvent or sample-sample interactions or the structure of solvent (water). They may stabilize or increase conformity of crystals by specific interactions with biological macromolecules.
The additive kit provides 48 different solutions of various additives, including organic solvents, salts, amino acids and other compounds. This kit provides an easy tool for rapid screening of additives. The kit includes 100 µl of each pre-formulated compound (solutions of organic solvent 1 mL).
Product #  Product Name Size
78374 Additive Kit 1 kit
Additive Kit Composition
Detergent Kit
Similar to the additive kit this kit provides a range of detergents that may be used for optimization. Detergents often interfere with the crystallization process. They are used to influence hydrophobic sample-sample interactions that may cause non-specific aggregation. On the other hand they can also influence the structure of water. The use of detergents is typically required for crystallization of membrane proteins or other proteins, if non-specific aggregation is suspected.
The detergent kit includes 48 defined detergents that are pre-formulated for ease of use. The concentration of each detergent solution is adjusted to the 10-fold value of critical micelle concentration. The kit includes 100 µl of each pre-formulated detergent.
Product #  Product Name Size
66317 Detergent Kit 1 kit
Detergent Kit Composition
Additive Kit Composition
Kit Compound No. Product #  Additive
1 61941 Ethylene glycol 5 M
2 38262 2-Methyl-2,4-pentanediol 3 M
3 50447 (+/-) -1,3-Butanediol 4 M
4 68891 1,4-Butanediol 4 M
5 28428 1,6 Hexanediol 3 M
6 50788 2,5 Hexanediol 3 M
7 39206 1,2,3-Hexanetriol 1 M
8 50791 1,2,3-Heptanetriol Isomer H 1 M
9 67190 1,2,3-Heptanetriol Isomer T 1 M
10 56592 1,2,3-Octanetriol Isomer H 1 M
11 97596 1,2,3-Octanetriol Isomer T 1 M
12 50722 Ammonium sulfate 1 M
13 43568 Cesium chloride 1 M
14 44675 Potassium chloride 1 M
15 40113 Lithium chloride 1 M
16 16818 Sodium chloride 2 M
17 08536 Natrium thiocyanate 2 M
18 08756 Sodium iodide 1 M
19 67414 Sodium fluoride 0.5 M
20 55298 Barium chloride 0.1 M
21 53704 Cadmium chloride 0.1 M
22 00457 Calcium chloride 0.1 M
23 28811 Cobalt chloride 0.1 M
24 15862 Cu(II)-chloride 0.1 M
25 18865 Magnesium chloride 0.1 M
26 48795 Manganese(II)-chloride 0.1 M
27 69042 Strontium chloride 0.1 M
28 28962 Yttrium chloride 0.1 M
29 39059 Zinc chloride 0.1 M
30 05292 Spermidine 0.1 M
31 28481 Spermine tetrahydrochloride 0.1 M
32 08740 6-Aminohexanoic acid 1 M
33 08730 1,5-Diaminopentanedihydrochloride 1 M
34 67258 1,6-Diaminohexane 1 M
35 16426 1,8-Diaminooctane 1 M
36 11102 3-(1-Pyridinio)-1-propanesulfonate 2 M
37 63226 Benzamidinhydrochloride 1 M
38 49541 Xylitol 1 M
39 02724 Taurine 0.1 M
40 18472 βine 0.1 M
41 64961 Trimethylamine hydrochloride 0.1 M
42 50535 Hexamminecobalt trichloride 0.1 M
43 18252 Ethylenediamine tetracetic acid Sodium salt 0.1 M
44 14495 L-Cysteine 0.1 M
45 08591 Sodium azide 0.1 M
46 12146 Glycerol 3 M
47 67419 Glycine 1 M
48 07458 Glycyl-glycyl-glycine 0.3 M
back to top
Detergent Kit Composition
Kit Compound No. Product #  Detergent
1 50006 Hexyl-β-D-glucoside 200 mM
2 44615 Heptyl-β-D-glucoside 150 mM
3 18842 Octyl-β-D-glucoside 100 mM
4 30887 Octyl-α/β-glucoside 100 mM
5 11448 Nonyl-β-D-glucoside 50 mM
6 04978 Methyl-6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside 200 mM
7 42387 Triethyleneglycol monoocyl ether 1 mM
8 41957 Triethyleneglycol monodecyl ether 1 mM
9 07891 Triethyleneglycol monododecyl ether 1 mM
10 13172 Tetraethylenglykolmonooctylether 1 mM
11 07888 Tetraethyleneglycol monodecyl ether 1 mM
12 16802 Tetraethyleneglycol monododecyl ether 1 mM
13 28000 Pentaethyleneglycol monodecyl ether 1 mM
14 04556 Pentaethyleneglycol monododecyl ether 1 mM
15 03269 Hexaethyleneglycol monodecyl ether 1 mM
16 15931 Hexaethyleneglycol monododecyl ether 1 mM
17 39376 Heptaethyleneglycol monodecyl ether 1 mM
18 49534 Heptaethyleneglycol monododecyl ether 1 mM
19 07511 Octaethyleneglycol monodecyl ether 1 mM
20 55377 Octaethyleneglycol monododecyl ether 1 mM
21 40913 Nonaethyleneglycol monododecyl ether 1 mM
22 07509 Decyl-β-D-maltoside 10 mM
23 04836 Undecyl-β-D-maltoside 10 mM
24 54202 Dodecyl-β-D-maltoside 3 mM
25 08386 Hexadecyl-β-D-maltoside 0.1 mM
26 54280 Cyclohexylmethyl-β-D-maltoside 500 mM
27 19164 2-Cyclohexylethyl-β-D-maltoside 200 mM
28 41583 6-Cyclohexylpentyl-β-D-maltoside 30 mM
29 51793 6-Cyclohexylhexyl-β-D-maltoside 30 mM
30 39489 Heptyl-β-D-thioglucoside 300 mM
31 07818 Nonyl-β-D-1-thiomaltopyranoside 100 mM
32 38498 Decyl-β-D-1-thiomaltopyranoside 100 mM
33 19899 CHAPS 100 mM
34 39490 MEGA-8 500 mM
35 44882 MEGA-9 200 mM
36 43104 N,N-Dimethylhexylamine N-oxide 100 mM
37 41261 N,N-Dimethyloctylamine N-oxide 100 mM
38 49556 N,N-Dimethylnonylamine N-oxide 100 mM
39 18705 N,N-Dimethyldecylamine N-oxide 100 mM
40 57915 N,N-Dimethyldodecylamine N-oxide 100 mM
41 54185 3-(N,N-Dimethyloctylammonio)propanesulfonate 500 mM
42 54196 3-(N,N-Dimethyldecylammonio)propanesulfonate 300 mM
43 16974 3-(N,N-Dimethyldodecylammonio)propanesulfonate 30 mM
44 53502 3-(N,N-Dimethylmyristoylammonio)propanesulfonate 3 mM
45 44490 3-(N,N-Dimethylpalmitoylammonio)propanesulfonate 0.3 mM
46 O-(n-Octyl)-phosphorylcholine 1 M
47 O-(n-Decyl)-phosphorylcholine 100 mM
48 O-(n-Tetradecyl)-phosphorylcholine 1.5 M
back to top
back to X-Ray Crystallography