Proteomics and Protein Expr.
Protein Expression
Cloning and Expression
Cell Lysis
Protocols
Purification and Detection
Protein Analysis
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 CelLytic IB for Solubilization of Inclusion Bodies
Bacterial Cell Lysis
CelLytic IB was designed to solubilize protein aggregates called inclusion bodies (Figure 1). In bacteria, inclusion bodies are sometimes formed when recombinant proteins are overexpressed. CelLytic IB was formulated to solubilize the protein of interest for immediate analysis of protein content or refolding procedures.
Product #  Product Name Package Size
C5236 CelLytic IB Cell Lysis Reagent 25 ml
100 ml
Figure 1. Solubilization of Streptavidin Inclusion Bodies.
Samples of solubilized inclusion body protein were assayed using BCA Reagent (BCA-1). 50 µl of each sample was incubated with BCA reagent at 60 °C for 15 min. The samples were then cooled to room temperature and assayed at 562 nm. The data above has been standardized to the protein recovery of CelLytic IB.
back to Bacterial Cell Lysis